Skip to Content
Merck
All Photos(1)

Documents

28-3160

Sigma-Aldrich

Sodium iodide

SAJ special grade, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaI
CAS Number:
Molecular Weight:
149.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ special grade

vapor density

>1 (vs air)

Assay

≥99.5%

form

crystalline

availability

available only in Japan

pH

6.0-9.0 (20 °C, 50 g/L)

mp

661 °C (lit.)

SMILES string

[Na+].[I-]

InChI

1S/HI.Na/h1H;/q;+1/p-1

InChI key

FVAUCKIRQBBSSJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brad A Bauer et al.
Physical chemistry chemical physics : PCCP, 14(6), 1892-1906 (2012-01-11)
We present results from all-atom molecular dynamics simulations of large-scale hydrophobic plates solvated in NaCl and NaI salt solutions. As observed in studies of ions at the air-water interface, the density of iodide near the water-plate interface is significantly enhanced
Caroline Serrano-Nascimento et al.
Molecular and cellular endocrinology, 349(2), 154-161 (2011-10-18)
Iodide excess acutely downregulates NIS mRNA expression, as already demonstrated. PCCl3 cells treated or not with NaI, NaI+NaClO(4) or NaI+Methimazole, for 30 min to 24 h, were used to further explore how iodide reduces NIS gene expression. NIS mRNA expression
Marlein Miranda Cona et al.
International journal of oncology, 44(3), 819-829 (2013-12-25)
Iodine-131‑labeled monoiodohypericin (131I‑Hyp) is a necrosis avid compound used as a complementary anticancer agent. Herein, the biodistribution in rats with re-perfused partial liver infarction (RPLI) was used to estimate its human internal radiation dosimetry. Iodine-123‑labeled monoiodohypericin (123I-Hyp) as a safer
Lan-Tao Li et al.
Chemical communications (Cambridge, England), 48(42), 5187-5189 (2012-04-20)
nBu(4)NI-catalyzed C3-selective formylation of N-H and N-substituted indoles by using N-methylaniline as a formylating reagent was first successfully demonstrated.
Yayoi S Kikkawa et al.
Journal of translational medicine, 12, 280-280 (2014-10-05)
A cochlear implant (CI) is an artificial hearing device that can replace a damaged cochlea. The present study examined the use of growth factor-eluting gelatin hydrogel coatings on the electrodes to minimize inner ear trauma during electrode insertion. Insulin-like growth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service