Skip to Content
Merck
All Photos(1)

Key Documents

28-2020

Sigma-Aldrich

Sodium borohydride

SAJ first grade, ≥95.0%

Synonym(s):

Sodium tetrahydridoborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBH4
CAS Number:
Molecular Weight:
37.83
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥95.0%
grade:
SAJ first grade
form:
solid
Pricing and availability is not currently available.

grade

SAJ first grade

Assay

≥95.0%

form

solid

reaction suitability

reagent type: reductant

availability

available only in Japan

mp

>300 °C (dec.) (lit.)

storage temp.

15-25°C

SMILES string

[BH4-].[Na+]

InChI

1S/BH4.Na/h1H4;/q-1;+1

InChI key

YOQDYZUWIQVZSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qinghua Cui et al.
Analytica chimica acta, 724, 86-91 (2012-04-10)
Silver nanoclusters (Ag NCs) templated with DNAs have attracted much attention as novel fluorophores because of their convenient emission tunability by the sequence and length of the template DNAs. However, the precise production of Ag NCs in a site-specific manner
Sylvie Girard et al.
American journal of reproductive immunology (New York, N.Y. : 1989), 72(4), 422-434 (2014-05-29)
Inflammation during pregnancy has devastating consequences for the placenta and fetus. These events are incompletely understood, thereby hampering screening and treatment. The inflammatory profile of villous tissue was studied in pregnancies at high-risk of placental dysfunction and compared to uncomplicated
Meganne L Christian et al.
ACS nano, 6(9), 7739-7751 (2012-08-10)
Owing to its high storage capacity (10.8 mass %), sodium borohydride (NaBH(4)) is a promising hydrogen storage material. However, the temperature for hydrogen release is high (>500 °C), and reversibility of the release is unachievable under reasonable conditions. Herein, we
Aldwin Suryo Rahmanto et al.
Free radical biology & medicine, 53(6), 1308-1316 (2012-08-14)
Singlet oxygen ((1)O(2)) is a reactive oxygen species generated during photo-oxidation, inflammation, and via peroxidase-catalyzed reactions (e.g., myeloperoxidase and eosinophil peroxidase). (1)O(2) oxidizes the free amino acids Trp, Tyr, His, Cys, and Met, and those species present on peptides/proteins, with
Marina Baccarin et al.
Materials science & engineering. C, Materials for biological applications, 58, 97-102 (2015-10-20)
A new architecture for a biosensor is proposed using a glassy carbon electrode (GCE) modified with hemoglobin (Hb) and silver nanoparticles (AgNPs) encapsulated in poly(amidoamine) dendrimer (PAMAM). The biosensors were characterized using ultraviolet-visible spectroscopy, ζ-potential and cyclic voltammetry to investigate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service