Skip to Content
Merck
All Photos(1)

Key Documents

28-1570

Sigma-Aldrich

Sodium acetate trihydrate

SAJ first grade, ≥98.5%

Synonym(s):

Acetic acid sodium salt trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
Beilstein:
3732037
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

SAJ first grade

Assay

≥98.5%

form

solid

autoignition temp.

1112 °F

availability

available only in Japan

pH

8.5-10 (25 °C, 408 g/L)

storage temp.

15-25°C

SMILES string

O.O.O.[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na.3H2O/c1-2(3)4;;;;/h1H3,(H,3,4);;3*1H2/q;+1;;;/p-1

InChI key

AYRVGWHSXIMRAB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Enzyme substrate for acetate kinase (EC 2.7.2.1)

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Horng-Ruey Chua et al.
Journal of critical care, 27(2), 138-145 (2012-03-24)
The purpose of the study was to determine the effects of Plasma-Lyte 148 (PL) vs 0.9% saline (NS) fluid resuscitation in diabetic ketoacidosis (DKA). A multicenter retrospective analysis of adults admitted for DKA to the intensive care unit, who received
Rishiram Ramanan et al.
FEBS letters, 587(4), 370-377 (2013-01-15)
Phenotypic and genotypic changes in Chlamydomonas reinhardtii BafJ5, a starchless mutant, with respect to lipid metabolism was studied in different trophic states under nitrogen (N) sufficient and limited conditions. Interestingly, cellular lipid content increased linearly with input acetate concentration with
Claire E Mason et al.
Journal of the American Society for Mass Spectrometry, 24(2), 274-285 (2013-01-04)
Single-stranded DNA-binding proteins (SSBs) are ubiquitous oligomeric proteins that bind with very high affinity to single-stranded DNA and have a variety of essential roles in DNA metabolism. Nanoelectrospray ionization mass spectrometry (nanoESI-MS) was used to monitor subunit exchange in full-length
Andrew D Shaw et al.
Annals of surgery, 255(5), 821-829 (2012-04-04)
To assess the association of 0.9% saline use versus a calcium-free physiologically balanced crystalloid solution with major morbidity and clinical resource use after abdominal surgery. 0.9% saline, which results in a hyperchloremic acidosis after infusion, is frequently used to replace
Qiong Jin et al.
Bioresource technology, 117, 148-154 (2012-05-23)
Dietzia natronolimnaea JQ-AN was isolated from industrial wastewater containing aniline. Under aerobic conditions, the JQ-AN strain degraded 87% of the aniline in a 300 mg L(-1) aniline solution after 120 h of shake flask incubation in a medium containing sodium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service