Skip to Content
Merck
All Photos(1)

Key Documents

28-1520

Sigma-Aldrich

Soda lime

for absorption of carbon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

form

solid

availability

available only in Japan

SMILES string

[OH-].[Na+].O=[Ca]

InChI

1S/Ca.Na.H2O.O/h;;1H2;/q;+1;;/p-1

InChI key

HUAUNKAZQWMVFY-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Caustic cocktail": closed-circuit diving apparatus contamination leading to corrosive injury.
Alicia B Minns et al.
The American journal of emergency medicine, 28(7), 843-843 (2010-09-15)
Satoshi Yagi et al.
Dental materials journal, 30(5), 611-615 (2011-09-29)
In this study, different glasses were investigated to improve reusable investments. Borosilicate glass (BSG) powder and soda-lime glass (SLG) powder were prepared by milling broken beakers and microscope slides, respectively, and used in experimental investments (I-BSG, I-SLG) by blending glass
Masahiro Shimizu et al.
Optics express, 20(2), 934-940 (2012-01-26)
We experimentally determined the three-dimensional temperature distribution and modification mechanism in a soda-lime-silicate glass under irradiation of ultrafast laser pulses at high repetition rates by analyzing the relationship between the morphology of the modification and ambient temperature. In contrast to
A Iu Elizarov et al.
Anesteziologiia i reanimatologiia, (2)(2), 8-10 (2011-06-22)
Using the mass spectrometric method we studied the interaction of volatile anesthetic sevoflurane with a CO2 absorber during low flow anesthesia (0.5 l/min fresh gas mixture). The results of measurements of sevoflurane and one of the most toxic breakdown products
Belén Cabal et al.
PloS one, 7(8), e42393-e42393 (2012-08-11)
This paper reports the effect of soda-lime-glass-nAg coating on the viability of an in vitro biofilm of Streptococcus oralis. Three strains (ATCC 35037 and two clinical isolates from periodontitis patients) were grown on coated with glass, glass containing silver nanoparticles

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service