Skip to Content
Merck
All Photos(1)

Key Documents

26-0020

Sigma-Aldrich

Sand, white quartz

CP, crystalline

Synonym(s):

Quartz, Silicon dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SiO2
CAS Number:
Molecular Weight:
60.08
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

grade

CP

form

crystalline

availability

available only in Japan

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

René Schneider et al.
Optics express, 21(3), 3523-3539 (2013-03-14)
Illumination based on objective-type total internal reflection (TIR) is nowadays widely used in high-performance fluorescence microscopy. However, the desirable application of such setups for dark-field imaging of scattering entities is cumbersome due to the spatial overlap of illumination and detection
Xujia Jiang et al.
Environmental pollution (Barking, Essex : 1987), 174, 38-49 (2012-12-19)
The significance of biofilm on the transport and deposition behaviors of ZnO nanoparticles were examined under a series of environmentally relevant ionic strength at two fluid velocities of 4 m-d(-1) and 8 m-d(-1). Biofilm enhanced nanoparticles retention in porous media
Lihui Feng et al.
Sensors (Basel, Switzerland), 13(9), 12482-12496 (2013-09-21)
A novel multi-fork z-axis gyroscope is presented in this paper. Different from traditional quartz gyroscopes, the lateral electrodes of the sense beam can be arranged in simple patterns; as a result, the fabrication is simplified. High sensitivity is achieved by
Sebastian Dochow et al.
Analytical and bioanalytical chemistry, 405(8), 2743-2746 (2013-02-02)
Three important technical innovations are reported here towards Raman-activated cell sorting. Firstly, a microfluidic chip made of quartz is introduced which integrates injection of single cells, trapping by laser fibres and sorting of cells. Secondly, a chip holder was designed
Peter Møller et al.
Critical reviews in toxicology, 43(2), 96-118 (2013-01-26)
Exposure to combustion-derived particles, quartz and asbestos is associated with increased levels of oxidized and mutagenic DNA lesions. The aim of this survey was to critically assess the measurements of oxidatively damaged DNA as marker of particle-induced genotoxicity in animal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service