24-3990
Potassium bicarbonate
JIS special grade, 99.5-100.3%
Synonym(s):
Potassium hydrogen carbonate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
KHCO3
CAS Number:
Molecular Weight:
100.12
Beilstein:
4535309
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
JIS special grade
form:
crystalline
Recommended Products
grade
JIS special grade
Assay
99.5-100.3%
form
crystalline
availability
available only in Japan
pH
8.2 (10.01 g/L)
SMILES string
[K+].OC([O-])=O
InChI
1S/CH2O3.K/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1
InChI key
TYJJADVDDVDEDZ-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Lisa Ceglia et al.
The Journal of clinical endocrinology and metabolism, 94(2), 645-653 (2008-12-04)
Protein is an essential component of muscle and bone. However, the acidic byproducts of protein metabolism may have a negative impact on the musculoskeletal system, particularly in older individuals with declining renal function. We sought to determine whether adding an
Karalee Jarvis et al.
Environmental science & technology, 43(16), 6314-6319 (2009-09-15)
Storage of CO2 through mineral sequestration using olivine has been shown to produce environmentally benign carbonates. However, due to the formation of a rate-limiting reaction product layer, the rate of reaction is insufficient for large-scale applications. We report the results
Fj Wegehaupt et al.
Oral diseases, 17(5), 508-514 (2011-02-22)
The aim of this study was to evaluate the erosive/abrasive enamel wear after contact with orange juices modified with different dietary supplements. A total of 96 bovine enamel samples were prepared and allocated to eight groups (1-8; n = 12).
K Holtenius et al.
Journal of dairy science, 91(2), 743-748 (2008-01-26)
The objective of the present study was to investigate the interactions of dietary K intake typical for forage-based diets on Mg balance in lactating dairy cows. Six lactating multiparous cows of the Swedish Red and White breed in midlactation were
Judith Buehlmeier et al.
The Journal of clinical endocrinology and metabolism, 97(12), 4789-4797 (2012-10-03)
High sodium chloride (NaCl) intake can induce low-grade metabolic acidosis (LGMA) and may thus influence bone and protein metabolism. We hypothesized that oral potassium bicarbonate (KHCO(3)) supplementation may compensate for NaCl-induced, LGMA-associated bone resorption and protein losses. Eight healthy male
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service