Skip to Content
Merck
All Photos(1)

Key Documents

24-3800

Sigma-Aldrich

Polyvinylpyrrolidone

CP, Mw 1200000, K 90, 11.5-12.8% N basis

Synonym(s):

PVP, Polyvidone, Povidone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H9NO)n
CAS Number:
MDL number:
UNSPSC Code:
12352005
grade:
CP

grade

CP

form

solid

mol wt

Mw 1200000

availability

available only in Japan

concentration

11.5-12.8% (N)

SMILES string

C=CN1CCCC1=O

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Polyvinylpyrrolidone is a component of Denhardt′s Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole M Sanders et al.
American journal of physiology. Endocrinology and metabolism, 294(5), E853-E860 (2008-03-13)
Selective serotonin reuptake inhibitors (SSRIs) are widely prescribed for patients with comorbid diabetes and depression. Clinical case studies in diabetic patients, however, suggest that SSRI therapy may exacerbate hypoglycemia. We hypothesized that SSRIs might increase the risk of hypoglycemia by
Chun-Mei Zhao et al.
Environmental toxicology and chemistry, 32(4), 913-919 (2013-01-25)
The toxicity of manufactured silver nanoparticles (AgNPs) has been widely studied, but the influence of AgNPs on the major ions (such as sodium [Na] and calcium [Ca]) regulations are unknown. In the present study, a freshwater cladoceran Daphnia magna was
S Fardindoost et al.
Nanotechnology, 24(13), 135201-135201 (2013-03-13)
In this paper we present experimental results describing electrical readout of the mechanical vibratory response of graphene-doped fibers by employing electrical actuation. For a fiber resonator with an approximate radius of 850 nm and length of 100 μm, we observed a resonance
Massimiliano di Cagno et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 48(4-5), 775-780 (2013-01-29)
The objective of this study was to investigate if the increase in apparent solubility induced by liposomalization or micellization of the poorly soluble drug hydrocortisone (HC) would lead to an enhancement of its permeability through biological membranes. For this purpose
Liandong Hu et al.
Pakistan journal of pharmaceutical sciences, 26(3), 629-636 (2013-04-30)
Solid dispersion technique has been developed many years for improving solubility of water-insoluble drugs, aiming to achieve a better oral bioavailability. However, this technique exhibits many inconveniences when used for large-scale tableting procedures. The objective of current research work was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service