Skip to Content
Merck
All Photos(1)

Key Documents

21-1690

Sigma-Aldrich

Nitric acid

JIS special grade, ≥97.0%, fuming, density: 1.52

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
Pricing and availability is not currently available.

grade

JIS special grade

vapor pressure

8 mmHg ( 20 °C)

Assay

≥97.0%

form

liquid

availability

available only in Japan

pH

<1 (20 °C)

pKa 

-1.3

bp

120.5 °C (lit.)

density

1.52

SMILES string

O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren E Kremer et al.
Journal of inorganic biochemistry, 147, 227-234 (2015-05-11)
Uncontrolled reactions in biological media are a main obstacle for clinical translation of V-based anti-diabetic or anti-cancer pro-drugs. We investigated the use of controlled-release pharmaceutical formulations to ameliorate this issue with a series of V(V) and (IV) complexes of anionic
Nicole McGowan et al.
The Science of the total environment, 494-495, 177-186 (2014-07-22)
Microchemical analysis of otolith (calcified 'ear stones' used for balance and orientation) of fishes is an important tool for studying their environmental history and management. However, the spatial resolution achieved is often too coarse to examine short-term events occurring in
Alyson M Baergen et al.
Environmental science & technology, 47(2), 815-820 (2012-12-15)
The fate of NO(x) (=NO + NO(2)) is important to understand because NO(x) is a significant player in air quality determination through its role in O(3) formation. Here we show that renoxification of the urban atmosphere may occur through the
Stijn Smulders et al.
Toxicological sciences : an official journal of the Society of Toxicology, 141(1), 132-140 (2014-06-14)
The unique physical and chemical properties of nanomaterials have led to their increased use in many industrial applications, including as a paint additive. For example, titanium dioxide (TiO2) engineered nanoparticles (ENPs) have well-established anti-UV, self-cleaning, and air purification effects. Silver
Peng Lei et al.
Molecular neurodegeneration, 9, 29-29 (2014-08-16)
We recently reported that Parkinsonian and dementia phenotypes emerge between 7-12 months of age in tau-/- mice on a Bl6/129sv mixed background. These observations were partially replicated by another group using pure Bl6 background tau-/- mice, but notably they did

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service