Skip to Content
Merck
All Photos(1)

Key Documents

19-0880

Sigma-Aldrich

Manganese(II) chloride tetrahydrate

SAJ first grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnCl2 · 4H2O
CAS Number:
Molecular Weight:
197.91
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

SAJ first grade

Assay

≥98.0%

form

flakes

availability

available only in Japan

pH

3.5-6 (25 °C, 50 g/L)

mp

58 °C (lit.)

SMILES string

Cl[Mn]Cl.[H]O[H].[H]O[H].[H]O[H].[H]O[H]

InChI

1S/2ClH.Mn.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2

InChI key

CNFDGXZLMLFIJV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - STOT RE 2

Target Organs

Brain

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seong-Uk Jin et al.
Magnetic resonance imaging, 31(7), 1143-1149 (2013-05-11)
The cochlear plays a vital role in the sense and sensitivity of hearing; however, there is currently a lack of knowledge regarding the relationships between mechanical transduction of sound at different intensities and frequencies in the cochlear and the neurochemical
Lisha Luo et al.
NMR in biomedicine, 25(12), 1360-1368 (2012-05-11)
The aim of this study was to provide data on the dose dependence of manganese-enhanced MRI (MEMRI) in the visual pathway of experimental rats and to study the toxicity of MnCl₂ to the retina. Sprague-Dawley rats were intravitreally injected with
Marta Sidoryk-Wegrzynowicz et al.
Journal of neurochemistry, 122(4), 856-867 (2012-06-20)
Manganese (Mn) has been implicated in the impairment of the glutamate-glutamine cycling (GGC) by deregulation of Glu and glutamine (Gln) turnover in astrocytes. Here, we have examined possible mechanisms involved in the Mn(II)-mediated disruption of Glu turnover, including those related
Geoffrey J Topping et al.
Medical physics, 40(4), 042502-042502 (2013-04-06)
Manganese(II) is employed as a contrast agent with magnetic resonance imaging (MRI) for study of neuronal activation in rats and mice. However, at the concentrations required for MRI, Mn may induce pharmacological or toxic effects. Positron emission tomography (PET) imaging
Anne Bertrand et al.
NeuroImage, 64, 693-702 (2012-09-11)
The impairment of axonal transport by overexpression or hyperphosphorylation of tau is well documented for in vitro conditions; however, only a few studies on this phenomenon have been conducted in vivo, using invasive procedures, and with contradictory results. Here we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service