Skip to Content
Merck
All Photos(1)

Key Documents

18-0800

Sigma-Aldrich

Lead(II) nitrate

SAJ first grade, ≥99.0%

Synonym(s):

Lead dinitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pb(NO3)2
CAS Number:
Molecular Weight:
331.21
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
SAJ first grade
form:
solid

grade

SAJ first grade

Assay

≥99.0%

form

solid

reaction suitability

reagent type: catalyst
core: lead

availability

available only in Japan

mp

470 °C (dec.) (lit.)

SMILES string

[PbH2++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/2NO3.Pb/c2*2-1(3)4;/q2*-1;+2

InChI key

RLJMLMKIBZAXJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Several new polymeric complexes of lead with dialkylamino derivatives of squaric acid were produced to study their coordination modes as possible prototypes for studies of lanthanide complexes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Repr. 1A - Skin Sens. 1B - STOT RE 1

Target Organs

Blood,Central nervous system,Immune system,Kidney

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao Shu et al.
Environmental science and pollution research international, 19(3), 893-902 (2011-10-04)
Cuttings and seedlings of Jatropha curcas L. were exposed to different regimes of lead (Pb) stress as Pb(NO₃)₂ at 0 (CK), 0.5, 1, 2, 3, and 4 mM kg⁻¹ soil. The effect of Pb treatment on the root length, tolerance
Kiyomitsu Nemoto et al.
The Journal of toxicological sciences, 36(3), 357-364 (2011-06-02)
Administration of lead ion (Pb) to rats and mice affects hepatic functions such as the induction of hepatic cell proliferation and upregulation of cholesterol biosynthesis. To identify the genes for which expression changes in response to Pb-administration, we analyzed hepatic
Kilarkaje Narayana et al.
Drug and chemical toxicology, 35(2), 208-217 (2011-10-13)
The aim of the present study was to investigate the coexistence of oxidative DNA damage and apoptosis- and necrosis-related DNA damage, and to correlate with ultrastructural changes in hepatocyte nuclei in the lead-nitrate-exposed liver. Adult male Wistar rats were exposed
M M Naik et al.
Letters in applied microbiology, 56(2), 99-104 (2012-11-21)
A lead-resistant bacteria isolated from soil contaminated with car battery waste were identified as Providencia alcalifaciens based on biochemical characteristics, FAME profile and 16S rRNA sequencing and designated as strain 2EA. It resists lead nitrate up to 0·0014 mol l(-1)
Mostafa Lamhamdi et al.
Comptes rendus biologies, 334(2), 118-126 (2011-02-22)
Lead (Pb) is an environmental pollutant extremely toxic to plants and other living organisms including humans. To assess Pb phytotoxicity, experiments focusing on germination of wheat seeds were germinated in a solution containing Pb (NO(3))(2) (0.05; 0.1; 0.5; 1g/L) during

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service