Skip to Content
Merck
All Photos(1)

Documents

13-2510

Sigma-Aldrich

8-Quinolinol

JIS special grade, ≥99.0%

Synonym(s):

8-Hydroxyquinoline, 8-Oxychinolin, Oxine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7NO
CAS Number:
Molecular Weight:
145.16
Beilstein:
114512
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.0%

form

solid

availability

available only in Japan

mp

72.5-74.0 °C

antibiotic activity spectrum

fungi

Mode of action

DNA synthesis | interferes
enzyme | inhibits

SMILES string

Oc1cccc2cccnc12

InChI

1S/C9H7NO/c11-8-5-1-3-7-4-2-6-10-9(7)8/h1-6,11H

InChI key

MCJGNVYPOGVAJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: quinolone

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David M Rubush et al.
Journal of the American Chemical Society, 134(33), 13554-13557 (2012-08-09)
The desymmetrization of p-peroxyquinols using a Brønsted acid-catalyzed acetalization/oxa-Michael cascade was achieved in high yields and selectivities for a variety of aliphatic and aryl aldehydes. Mechanistic studies suggest that the reaction proceeds through a dynamic kinetic resolution of the peroxy
Bhimashankar Mitkari et al.
Experimental neurology, 239, 158-162 (2012-10-13)
Cell therapies from various sources have been under intense research in stroke. Efficient homing of the cells to the injured brain without complications is necessary to realize the therapeutic potential of cell therapy. Intra-arterial (IA) infusion of cells bypasses the
M Luísa Ramos et al.
Dalton transactions (Cambridge, England : 2003), 41(40), 12478-12489 (2012-09-08)
Multinuclear ((1)H, (13)C and (27)Al) magnetic resonance spectroscopy (1D and 2D), DFT calculations and fluorescence have been used to study the complexation of 8-hydroxyquinoline-5-sulfonate (8-HQS) with Al(III). The study combines the high sensitivity of luminescence techniques, the selectivity of multinuclear
Steve Comby et al.
Inorganic chemistry, 51(19), 10158-10168 (2012-09-15)
A novel near-infrared (NIR) emissive lanthanide-based zinc sensor was designed, based on the self-assembly in aqueous solution between the nonemissive coordinatively unsaturated Yb(III) cyclen complex 2·Yb and the sulfonated 8-hydroxyquinoline (8-HQS) chromophore, which was employed as a sensitizing antenna. The
G L Apostoli et al.
Journal of thrombosis and haemostasis : JTH, 12(11), 1880-1889 (2014-08-29)
Nitric oxide (NO) is a critical negative regulator of platelets that is implicated in the pathology of thrombotic diseases. Platelets generate NO, but the presence and functional significance of NO synthase (NOS) in platelets is unclear. Inorganic nitrate/nitrite is increasingly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service