Skip to Content
Merck
All Photos(1)

Key Documents

09-1400

Sigma-Aldrich

Ethylenediaminetetraacetic acid disodium magnesium salt tetrahydrate

SAJ special grade, ≥99.0%

Synonym(s):

Ethylenediaminetetraacetic acid disodium magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NaOOCCH2)2NCH2CH2N(CH2COO)2Mg · 4H2O
CAS Number:
Molecular Weight:
430.56
EC Number:
MDL number:
UNSPSC Code:
12352106
Pricing and availability is not currently available.

grade

SAJ special grade

Assay

≥99.0%

availability

available only in Japan

SMILES string

O=C(CN(CCN(CC(O[Na])=O)CC(O[Na])=O)C1)O[Mg]OC1=O.C

InChI

1S/C10H16N2O8.CH4.Mg.2Na/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);1H4;;;/q;;+2;2*+1/p-4

InChI key

GWHGOZUTJQPIGV-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service