Skip to Content
Merck
All Photos(1)

Documents

05-1200

Sigma-Aldrich

(±)-Camphor

SAJ first grade, ≥96.0%

Synonym(s):

1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
1907611
EC Number:
MDL number:
UNSPSC Code:
12352212
PubChem Substance ID:

grade

SAJ first grade

vapor density

5.2 (vs air)

vapor pressure

4 mmHg ( 70 °C)

Assay

≥96.0%

expl. lim.

3.5 %

availability

available only in Japan

bp

204 °C (lit.)

mp

175-177 °C (lit.)

SMILES string

[H][C@](CC1=O)(CC2)C(C)(C)[C@]12C

InChI

1S/C10H16O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7H,4-6H2,1-3H3/t7-,10+/m1/s1

InChI key

DSSYKIVIOFKYAU-XCBNKYQSSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 2 Inhalation

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

147.9 °F - closed cup

Flash Point(C)

64.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon P Skinner et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(29), 9022-9027 (2015-07-02)
The energy landscapes of proteins are highly complex and can be influenced by changes in physical and chemical conditions under which the protein is studied. The redox enzyme cytochrome P450cam undergoes a multistep catalytic cycle wherein two electrons are transferred
Hiroyuki Kitahata et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 87(1), 010901-010901 (2013-02-16)
The coupling between deformation and motion in a self-propelled system has attracted broader interest. In the present study, we consider an elliptic camphor particle for investigating the effect of particle shape on spontaneous motion. It is concluded that the symmetric
Christian Merten et al.
The journal of physical chemistry. A, 116(27), 7329-7336 (2012-06-06)
In the present work, the first observation of strong resonance Raman optical activity (RROA) involving more that one resonant electronic state is reported. The chiral transition metal complex bis-(trifluoroacetylcamphorato) copper(II), abbreviated Cu(tfc)(2), exhibits both resonance Raman (RR) and RROA spectra
Yali V Zhang et al.
Nature neuroscience, 16(10), 1468-1476 (2013-09-10)
Animals tend to reject bitter foods. However, long-term exposure to some unpalatable tastants increases acceptance of these foods. Here we show that dietary exposure to an unappealing but safe additive, camphor, caused the fruit fly Drosophila melanogaster to decrease camphor
Hiroyuki Yamashita et al.
Pharmaceutical research, 30(1), 70-80 (2012-08-22)
Although a number of studies have reported that cocrystals can form by heating a physical mixture of two components, details surrounding heat-induced cocrystal formation remain unclear. Here, we attempted to clarify the thermal behavior of a physical mixture and cocrystal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service