Skip to Content
Merck
All Photos(1)

Key Documents

05-0670

Sigma-Aldrich

Calcium fluoride

SAJ first grade, ≥97.0%

Synonym(s):

Fluorite, Fluorspar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaF2
CAS Number:
Molecular Weight:
78.07
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
SAJ first grade
form:
powder
Pricing and availability is not currently available.

grade

SAJ first grade

Assay

≥97.0%

form

powder

availability

available only in Japan

bp

2500 °C (lit.)

density

3.18 g/mL at 25 °C (lit.)

SMILES string

F[Ca]F

InChI

1S/Ca.2FH/h;2*1H/q+2;;/p-2

InChI key

WUKWITHWXAAZEY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason S Lupoi et al.
Biotechnology for biofuels, 8, 98-98 (2015-07-23)
Slow-degrading, fossil fuel-derived plastics can have deleterious effects on the environment, especially marine ecosystems. The production of bio-based, biodegradable plastics from or in plants can assist in supplanting those manufactured using fossil fuels. Polyhydroxybutyrate (PHB) is one such biodegradable polyester
G Rølla
Acta odontologica Scandinavica, 46(6), 341-345 (1988-12-01)
The literature concerning the formation and stability of CaF2 in the oral environment is reviewed. In early work the CaF2 formed during topical application with fluoride was assumed to be beneficial. It was suggested that it could protect the enamel
Adchara Padermshoke et al.
Applied spectroscopy, 66(6), 711-718 (2012-06-27)
A novel SiO(2)-deposited CaF(2) (SiO(2)/CaF(2)) substrate for measuring vibrational sum-frequency generation (SFG) spectra of silane-based chemisorbed monolayers in aqueous media has been developed. The substrate is suitable for silanization and transparent over a broad range of the infrared (IR) probe.
Yunlu Dai et al.
Biomaterials, 33(8), 2583-2592 (2011-12-27)
In this study, we design a controlled release system based on CaF(2):Ce(3+)/Tb(3+)-poly(acrylic acid) (PAA) composite microspheres, which were fabricated by filling the pH-responsive PAA inside CaF(2):Ce(3+)/Tb(3+) hollow spheres via photopolymerization route. The CaF(2):Ce(3+)/Tb(3+) hollow spheres prepared by hydrothermal route possess
Willem Van den Heuvel et al.
Physical review letters, 109(7), 073001-073001 (2012-09-26)
We present a theory of nuclear magnetic resonance shielding tensors for electronic states with arbitrary degeneracy. The shieldings are here expressed in terms of generalized Zeeman (g((k))) and hyperfine (A((k))) tensors, of all ranks k allowed by the size of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service