Skip to Content
Merck
All Photos(1)

Key Documents

01-3470

Sigma-Aldrich

4-Aminobenzoic acid

98.5-100.2%, SAJ first grade

Synonym(s):

PABA, Vitamin Bx, Vitamin H1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CO2H
CAS Number:
Molecular Weight:
137.14
Beilstein:
471605
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

SAJ first grade

Assay

98.5-100.2%

form

solid

availability

available only in Japan

mp

186-189 °C
187-189 °C (lit.)

solubility

water: 4.7 g/L at 20 °C

density

1.374 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

Nc1ccc(cc1)C(O)=O

InChI

1S/C7H7NO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,8H2,(H,9,10)

InChI key

ALYNCZNDIQEVRV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

339.8 °F - closed cup

Flash Point(C)

171 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L O Derewlany et al.
The Journal of pharmacology and experimental therapeutics, 269(2), 756-760 (1994-05-01)
The fetus is exposed to almost all of the substances found in the maternal circulation whether nutrients or foreign chemicals ("xenobiotics"). The main route of exposure is the placenta. The placenta is metabolically active toward xenobiotics and the nature of
L O Derewlany et al.
The Journal of pharmacology and experimental therapeutics, 269(2), 761-765 (1994-05-01)
Studies in our laboratory have shown that the N-acetylation activity of the human term placenta is a predominantly attributable to the NAT1 form of arylamine N-acetyltransferase (NAT). To further assess the acetylation capacity of the placenta, the N-acetylation of the
Shu-Ting Chen et al.
Journal of the American Society for Mass Spectrometry, 23(8), 1408-1418 (2012-06-08)
A strategy based on negative ion electrospray ionization tandem mass spectrometry and closed-ring labeling with both 8-aminopyrene-1,3,6-trisulfonate (APTS) and p-aminobenzoic acid ethyl ester (ABEE) was developed for linkage and branch determination of high-mannose oligosaccharides. X-type cross-ring fragment ions obtained from
Yoshihiko Tashiro et al.
Blood, 119(26), 6382-6393 (2012-05-11)
Plasminogen activator inhibitor-1 (PAI-1), an endogenous inhibitor of a major fibrinolytic factor, tissue-type plasminogen activator, can both promote and inhibit angiogenesis. However, the physiologic role and the precise mechanisms underlying the angiogenic effects of PAI-1 remain unclear. In the present
Lei Zhou et al.
Water research, 47(1), 153-162 (2012-10-23)
Dissolved organic matter (DOM) is an important photosensitizer for the phototransformation of organic contaminants in sunlit natural waters. This article focuses on the photolysis kinetics and mechanism of sunscreen agent p-aminobenzoic acid (PABA) in the presence of four kinds of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service