Skip to Content
Merck
All Photos(1)

Key Documents

01-3085

Sigma-Aldrich

Aluminum sulfate hexadecahydrate

SAJ first grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2(SO4)3 · 16H2O
CAS Number:
Molecular Weight:
630.40
EC Number:
MDL number:
UNSPSC Code:
12352311
PubChem Substance ID:

grade

SAJ first grade

form

solid

availability

available only in Japan

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Al.3H2O4S.16H2O/c;;3*1-5(2,3)4;;;;;;;;;;;;;;;;/h;;3*(H2,1,2,3,4);16*1H2/q2*+3;;;;;;;;;;;;;;;;;;;/p-6

InChI key

TYYRFZAVEXQXSN-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nagasathiya Krishnan et al.
European journal of mass spectrometry (Chichester, England), 21(3), 481-486 (2015-08-27)
The coordinating nature of the hydroxy carboxylic acids, such as tartaric and citric acids, has been utilized for the in-situ formation of anions representing the trivalent elements boron and aluminum and two dianions of the hydroxy acid selected under negative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service