Skip to Content
Merck
All Photos(1)

Key Documents

01-2740

Sigma-Aldrich

Aluminum hydroxide

CP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(OH)3
CAS Number:
Molecular Weight:
78.00
EC Number:
MDL number:
UNSPSC Code:
12352311
PubChem Substance ID:
Pricing and availability is not currently available.

grade

CP

form

solid

availability

available only in Japan

solubility

water: insoluble

density

2.42 g/cm3 at 20 °C

SMILES string

O[Al](O)O

InChI

1S/Al.3H2O/h;3*1H2/q+3;;;/p-3

InChI key

WNROFYMDJYEPJX-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne Szarewski et al.
The Journal of infectious diseases, 208(9), 1391-1396 (2013-10-05)
Public Health England has reported a decrease of up to 20.8% in new diagnoses of external genital warts (GWs) among women aged <19 years since the national vaccination program with the human papillomavirus (HPV)-16/18 AS04-adjuvanted vaccine began in 2008. A
Micha Phill Grønholm Jepsen et al.
The Journal of infectious diseases, 208(3), 479-488 (2013-04-30)
GMZ2 is a hybrid protein consisting of the N-terminal region of the glutamate-rich protein fused in frame to the C-terminal region of merozoite surface protein 3 (MSP3). GMZ2 formulated in Al(OH)3 has been tested in 3 published phase 1 clinical
Wolfgang Hoehenwarter et al.
Molecular & cellular proteomics : MCP, 12(2), 369-380 (2012-11-23)
Mitogen-activated protein kinase (MPK) cascades are important for eukaryotic signal transduction. They convert extracellular stimuli (e.g. some hormones, growth factors, cytokines, microbe- or damage-associated molecular patterns) into intracellular responses while at the same time amplifying the transmitting signal. By doing
Masaoki Kimura et al.
Water research, 47(6), 2075-2084 (2013-02-21)
Aluminum coagulants are widely used in water treatment plants to remove turbidity and dissolved substances. However, because high aluminum concentrations in treated water are associated with increased turbidity and because aluminum exerts undeniable human health effects, its concentration should be
Jia Wang et al.
Journal of ethnopharmacology, 154(1), 131-147 (2014-04-08)
The prevalence of allergic asthma has been increased rapidly in recent years. About 20% of all these sufferers have experienced asthma exacerbation. Although corticosteroids and β-agonists therapy improves serious asthma symptoms, they can׳t completely cure these allergic diseases. BuShenYiQi Formula

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service