Skip to Content
Merck
All Photos(1)

Documents

NTMIXKB

Roche

KAPA dNTP Mix (10 mM each)

Synonym(s):

DNTP, PCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

grade

Molecular Biology

Quality Level

Assay

>99% (HPLC)

form

liquid

shelf life

≤18 mo.

packaging

pkg of 5 mL (KK1041)
pkg of 500 μL (KK1017)

manufacturer/tradename

Roche

impurities

Bacterial contamination assay, passes test
DNase, none detected
Endonuclease contamination assay, passes test
Exonuclease contamination assay, passes test
Human contamination assay, passes test
RNase, none detected
nicking activity, none detected
phosphatase, none detected
protease, none detected

color

colorless

solubility

water: miscible

shipped in

dry ice

storage temp.

−20°C

General description

KAPA dinucleotide triphosphates (dNTPs) provide high-quality, molecular, premium grade dinucleotide phosphates (dNTPs) mainly used for polymerase chain reaction (PCR).

Application

KAPA dNTP Mix has been used in:
  • Standard PCR
  • High fidelity PCR
  • Quantitative PCR
  • End-repair or A-tailing
  • cDNA synthesis
  • DNA sequencing
  • DNA labeling
  • Microarrays
KAPA dNTPs are suitable for use in any molecular biology application requiring high-quality dNTPs.

Features and Benefits

  • KAPA dNTPs are available as individual dNTPs or as a ready-to-use dNTP Mix.
  • KAPA dNTP Mix is supplied with KAPA Taq, KAPA2G Fast, KAPA2G Robust, KAPA LongRange and KAPA HiFi PCR Kits.
  • KAPA dNTPs are subjected to strict quality control to ensure high performance, particularly with B-family polymerases that are sensitive to poisoning by uracil.
  • Compatible with any commercially available DNA polymerases.

Quality

KAPA dNTPs are >99% pure by HPLC and are free of DNase, RNase, protease, phosphatase, and nicking activity. All kit components are subjected to stringent functional quality control, and meet strict requirements with respect to DNA contamination.

Preparation Note

Always ensure that the product KAPA dNTP Mix has been fully thawed and mixed before use. KAPA dNTPs may be stored at 4°C for short-term use (up to 1 month). Return to -20°C for long-term storage. Provided that the sample has been handled carefully and not contaminated, the samples are not expected to be compromised if left (unintentionally) at room temperature for a short period of time (up to 3 days). Long-term storage at room temperature and 4°C is not recommended. Please note that storing the reagents at temperatures above -20°C makes them more prone to degradation when contaminated during use, and therefore storage at such temperatures is at the user′s own risk.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Mixture of dATP, dCTP, dGTP and dTTP 10 mM each

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Distinct classes of chromatin loops revealed by deletion of an RNA-binding region in CTCF
Hansen AS, et al.
Molecular Cell, 76 (2019)
BRAF VE1 immunoreactivity patterns in epithelioid glioblastomas positive for BRAF V600E mutation.
Kleinschmidt-DeMasters B K, et al.
American Journal of Surgical Pathology, 39(4), 528-528 (2015)
Novel TP53 gene mutation and correlation with p53 immunohistochemistry in a mixed epithelial carcinoma of the endometrium.
Sholl A B, et al.
Gynecologic oncology case reports, 3, 11-11 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service