Skip to Content
Merck
All Photos(1)

Key Documents

11719394001

Roche

Immunoprecipitation Kit (Protein A)

sufficient for 20 reactions, kit of 1, suitable for immunoprecipitation (IP)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

usage

sufficient for 20 reactions

packaging

kit of 1

manufacturer/tradename

Roche

technique(s)

immunoprecipitation (IP): suitable

storage temp.

2-8°C

General description

Immunoprecipitation is a widely used method for the analysis of target antigens in complex mixture of proteins. The protein of interest can be concentrated and immunoaffinity -purified in one step on an analytical scale via a specific antibody. Often, immunoprecipitated proteins are functionally fully active and can be further analyzed with respect to enzymatic activity, interactions, modifications and structure.
Kit contains all reagents necessary for cell lysis, solubilization, stabilization, and immunopurification of proteins.

Application

Immunoprecipitation Kit (Protein A) has been used for the immunoprecipitation of proteins from cellular extracts with Protein A Agarose.[1][2][3]

Packaging

1 kit containing 5 components.

Preparation Note

Working solution: Lysis buffer/wash buffer 1
The kit contains reagents for 125 ml of lysis buffer/wash buffer 1. Prepare at least a minimal volume of 25 ml, sufficient for four immunoprecipitations.
To prepare 25 ml of lysis buffer/wash buffer 1 mix 5 ml core buffer, 3.75 ml NaCl, 2.5 ml detergent mix and 1 cOmplete tablet. Add water to a final volume of 25 ml.
Solution is stable at 2 to 8 °C for 24 hours. When stored in aliquots at -15 to -25 °C, the solution is stable for at least four weeks. Mix thoroughly after thawing.

Wash buffer 2
The kit contains reagents for 50 ml of wash buffer 2. 2 ml of this buffer is required for one immunoprecipitation.
To prepare 50 ml of wash buffer 2 mix 10 ml core buffer, 25 ml NaCl and 0.5 ml detergent mix. Add water to a final volume of 50 ml.
Solution is stable at 2 to 8 °C. For longer periods, store aliquots at -15 to -25 °C. Mix thoroughly after thawing.

Wash buffer 3
The kit contains reagents for 25 ml of wash buffer 3. 1 ml of this buffer is required for one immunoprecipitation.
To prepare 25 ml mix 1 ml core buffer and 0.25 ml detergent mix. Add water to a final volume of 25 ml.
Solution is stable at 2 to 8 °C. For longer periods, store aliquots at -15 to -25 °C. Mix thoroughly after thawing.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Core Buffer

  • NaCl

  • Detergent Mix

  • cOmplete Protease Inhibitor Cocktail Tablets (5)

  • Protein A Agarose ready-to-use

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dual-specificity phosphatase 9 Protects against nonalcoholic fatty liver disease in mice through ASK1 suppression
Ye P, et al.
Hepatology, 69(1), 76-93 (2019)
Yi Liu et al.
Cardiovascular research, 101(1), 87-96 (2013-11-29)
Toll-interacting protein (Tollip) is a critical regulator of the Toll-like receptor-mediated signalling pathway. However, the role of Tollip in chronic pressure overload-induced cardiac hypertrophy remains unclear. This study aimed to determine the functional significance of Tollip in the regulation of
J A Arnott et al.
Bone, 42(5), 871-885 (2008-03-04)
Connective tissue growth factor (CTGF/CCN2) is a cysteine rich, extracellular matrix protein that acts as an anabolic growth factor to regulate osteoblast differentiation and function. In osteoblasts, CTGF is induced by TGF-beta1 where it acts as a downstream mediator of
Ding-Sheng Jiang et al.
Hypertension (Dallas, Tex. : 1979), 63(4), 713-722 (2014-01-08)
Cardiac hypertrophy is a complex pathological process that involves multiple factors including inflammation and apoptosis. Interferon regulatory factor 7 (IRF7) is a multifunctional regulator that participates in immune regulation, cell differentiation, apoptosis, and oncogenesis. However, the role of IRF7 in
Shu-Min Zhang et al.
Molecular and cellular biology, 34(3), 400-414 (2013-11-20)
Interferon regulatory factor 8 (IRF8), a member of the IRF transcription factor family, was recently implicated in vascular diseases. In the present study, using the mouse left carotid artery wire injury model, we unexpectedly observed that the expression of IRF8

Protocols

Immunoprecipitation Kit (Protein A) Protocol & Troubleshooting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service