Skip to Content
Merck
All Photos(1)

Documents

11204530001

Roche

ABTS Buffer

solution, pkg of 125 mL

Synonym(s):

buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

form

solution

Quality Level

packaging

pkg of 125 mL

manufacturer/tradename

Roche

shipped in

wet ice

storage temp.

2-8°C

General description

2,2′-azino-di-(3-ethylbenzthiazoline sulfonic acid) or ABTS acts as a substrate for HRP (horseradish peroxidase) conjugate during enzyme-linked immunosorbent assay (ELISA). It is the most sensitive, and stable substrate when compared to three other substrates namely, 5-aminosalicylic acid (5AS), O-phenylenediamine (OPD), O-tolidine (OT). It also produces the best visual results, where it gives a bluish-green color. ELISA using ABTS is a highly sensitive, specific and reproducible technique.

Application

ABTS Buffer is a solvent for ABTS. ABTS solution is the ideal substrate solution for enzyme immunoassays with horseradish peroxidase (HRP) as a marker enzyme. Because of the high sensitivity of the reaction, autoreaders will often display "over" after several minutes.

Preparation Note

Working concentration: Flow cytometry: 0.5 μg/100 μl (106 cells); Immunohistocytochemistry: 50 μg/ml
Working concentration of conjugate depends on application and substrate. Concentrations should be taken as a guideline.
Working solution: Dissolve one ABTS tablet (5 mg) in 5 ml of ABTS buffer. This solution has a light-green color. In microtiter plates, the solution is colorless A405 nm/1 cm should be <0.16. Work as clean as possible. Heavy metal ions or traces of peroxidase can produce a dark green ABTS solution. Such solutions are not suitable for the assay.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

ABTS is a trademark of Roche

WGK

nwg

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Lindenthal et al.
Infection and immunity, 69(1), 52-57 (2000-12-19)
Enterotoxigenic Escherichia coli (ETEC) is capable of invading epithelial cell lines derived from the human ileum and colon. Two separate invasion loci (tia and tib) that direct noninvasive E. coli strains to adhere to and invade cultured human intestine epithelial
Patrik Erlmann et al.
Molecular biology of the cell, 20(20), 4400-4411 (2009-08-28)
Deleted in Liver Cancer 1 (DLC1) is a GTPase-activating protein (GAP) with specificity for RhoA, RhoB, and RhoC that is frequently deleted in various tumor types. By inactivating these small GTPases, DLC1 controls actin cytoskeletal remodeling and biological processes such
Bruce S Sachais et al.
Blood, 120(5), 1137-1142 (2012-05-12)
Rapid laboratory assessment of heparin-induced thrombocytopenia (HIT) is important for disease recognition and management. The utility of contemporary immunoassays to detect antiplatelet factor 4 (PF4)/heparin antibodies is hindered by detection of antibodies unassociated with disease. To begin to distinguish properties
H Matsuda et al.
The Japanese journal of experimental medicine, 54(3), 131-138 (1984-06-01)
A micro-technique of enzyme-linked immunosorbent assay (ELISA) using ABTS, 2,2'-azino-di-(3-ethylbenzthiazoline sulfonic acid), as a substrate for horseradish peroxidase (HRP) conjugate was studied. In a comparative study among 4 substrates, namely; 5-aminosalicylic acid (5AS), O-phenylenediamine (OPD), O-tolidine (OT) and ABTS, for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service