Skip to Content
Merck
All Photos(1)

Key Documents

11164368001

Roche

N-Acetyl-β-D-Glucosaminidase Control (NAG Control)

from beef kidney

Synonym(s):

NAG control

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
Pricing and availability is not currently available.

biological source

bovine kidney

Quality Level

form

lyophilized

specific activity

~9 mU/mL

packaging

pkg of 3 × 1 mL

manufacturer/tradename

Roche

technique(s)

activity assay: suitable

color

white to light yellow

solubility

water: soluble

suitability

suitable for enzyme test

UniProt accession no.

application(s)

life science and biopharma

storage temp.

2-8°C

General description

Control solution for the determination of NAG activity in urine research samples with the aid of the assay kit N-acetyl-β-D-glucosaminidase.

Application

Additional control solution for the determination of NAG activity in urine in life science research applications with the aid of N-acetyl-β-D-glucosaminidase (NAG) on automatic analyzers.

Unit Definition

Volume Activity: The activity of the NAG control is ca. 9 mU N-acetyl-β-D-glucosaminidase from beef kidney per ml. For exact value see data on the label.

Preparation Note

Working solution: Preparation of the standard solution
  • Dissolve contents of one bottle with 1 ml double-dist. water, close bottle and wait for 5 minutes.
  • Dissolve contents completely by gentle swirling.
    Note: Avoid foaming! The solution is stable for at least 1 month stored at 2 to 8 °C, protected from light.

Reconstitution

Preparation of the standard solution
  • Dissolve contents of one bottle with 1 ml double-dist. water, close bottle and wait for 5 minute.
  • Dissolve contents completely by gentle swirling.
    Note: Avoid foaming! The solution is stable for at least 1 month stored at 2 to 8 °C, protected from light.

Storage and Stability

protect from light.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service