Skip to Content
Merck
All Photos(2)

Documents

04743784001

Roche

GC-RICH PCR System, dNTPack

suitable for PCR, hotstart: no, Difficult Templates/Specialty Enzymes PCR

Synonym(s):

GC-rich PCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.21

usage

sufficient for ≤50 reactions

feature

Difficult Templates/Specialty Enzymes PCR
dNTPs included
hotstart: no

packaging

pkg of 100 U

manufacturer/tradename

Roche

parameter

72 °C optimum reaction temp.

technique(s)

PCR: suitable

input

purified DNA

shipped in

dry ice

storage temp.

−20°C (−15°C to −25°C)

Related Categories

General description

The GC-RICH PCR System, dNTPack is composed of a special enzyme blend of thermostable Taq DNA Polymerase and Tgo DNA Polymerase, a thermostable enzyme with a proofreading (3′-5′ exonuclease) activity. This polymerase mixture by itself outperforms Taq DNA Polymerase in respect to yields, fidelity and specificity beside the possibility to amplify fragments up to 5 kb in length. The GC-RICH PCR reaction buffer in combination with the included GC-RICH resolution solution allows to amplify very efficiently difficult templates like GC-rich targets.

Application

The GC-RICH PCR System, a blend of Taq DNA Polymerase and a proofreading polymerase, enables amplification of templates that are difficult or impossible to amplify with other polymerases and other blends of polymerases. The enhanced processivity of the blend and the unique GC-RICH Resolution Solution combine to deliver superior performance – especially from problematic templates.
The GC-RICH PCR System, dNTPack may also be used in standard PCR applications, providing improved results (higher yield, higher accuracy) over Taq DNA Polymerase alone.

Features and Benefits

  • Ease access to difficult templates:-
including GC-rich targets and repetitive sequences.
  • Reagents, the GC-RICH Resolution Solution and PCR Grade Water are provided.
  • Amplify DNA fragments up to 5 kb.
  • Cost-effective:-
use the convenient premixed solution of PCR grade NTPs.

Packaging

1 kit containing 6 components

Quality

The GC-RICH PCR System is function-tested, by amplifying a human 284 bp ApoE genomic fragment, using the GC-RICH PCR System protocols.

Unit Definition

Volume Activity: 2 U/μl

Preparation Note

Working concentration: The optimal enzyme concentration range from 0.5 to 5 U per assay. For a standard 50 μl PCR, we recommend using 2 U of the enzyme blend.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

Use of this product is covered by US patent claims and corresponding patent claims outside the US. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patent claims require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.

Kit Components Only

Product No.
Description

  • GC-RICH Enzyme Mix, in storage buffer

  • GC-RICH PCR Reaction Buffer, with 7.5 mM MgCl2 and DMSO 5x concentrated

  • GC-RICH Resolution Solution 5 M

  • MgCl2 Stock Solution 25 mM

  • Water, PCR Grade

  • PCR Nucleotide Mix

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Distribution of Giardia duodenalis assemblages A and B among children living in a remote indigenous community of the Northern Territory, Australia
Asher, et al.
PLoS ONE, 9 (2014)
Jenny Rinke et al.
Clinical chemistry, 59(8), 1238-1250 (2013-04-18)
We sought to establish a convenient, sensitive next-generation sequencing (NGS) method for genotyping the 26 most commonly mutated leukemia-associated genes in a single work flow and to optimize this method for low amounts of input template DNA. We designed 184
Chromothripsis Is a Recurrent Genomic Abnormality in High-Risk Myelodysplastic Syndromes
Abaigar, et al.
PLoS ONE, 11 (2016)
Laurianne Davignon et al.
Human molecular genetics, 25(8), 1559-1573 (2016-03-25)
Despite recent progress in the genetic characterization of congenital muscle diseases, the genes responsible for a significant proportion of cases remain unknown. We analysed two branches of a large consanguineous family in which four patients presented with a severe new

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service