Skip to Content
Merck
All Photos(1)

Documents

XX1604705

Millipore

Vacuum Flask, 2 L

Accessories for filter holders for sample preparation.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104909
eCl@ss:
32031613

material

borosilicate glass

manufacturer/tradename

MilliSolve

flask capacity

2 L

application(s)

filtration

shipped in

ambient

General description

Accessories for Glass Filter Holders – XX100900, XX1604705, XX1604706, XX1009010, XX1009002





The All-Glass Filter Holder was designed for the filtration of aqueous and organic or corrosive liquids for particulate contamination analysis. The holder can also be used for HPLC solvent preparation. The vacuum connection is integrated into the filter holder base and flask cap, above the filtrate exit level. With this design, you can avoid unintentionally drawing filtrate into vacuum tubing. This design also allows simple transfer of filtrate (i.e., HPLC solvents) by pouring from the receiver flask.



These part numbers are accessory items for the all glass filter holders.



Features & Benefits:

•All-glass construction has broad chemical compatibility

•Borosilicate glass parts contact liquid, with ground-glass sealing surfaces

•Vacuum connection to flask cap simplifies transfer of filtrate



Applications:

Particle Contamination Analysis, HPLC Solvent Filtration, General Filtration and Clarification

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service