Skip to Content
Merck
All Photos(2)

Key Documents

UFSC050MT

Millipore

Amicon® Stirred Cell 50mL Maintenance Kit (stir bar assembly, 2 each seals, tubing kit)

This Amicon stirred cell model provides high flow rates with solutions up to 10% macrosolute concentration. Capable of rapid concentration, or salt removal followed by concentration in the same unit.

Synonym(s):

50mL Stirred Cell Kit, Amicon Maintenance Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
eCl@ss:
32031690
NACRES:
NB.24
Pricing and availability is not currently available.

material

Nylon connector (Luer; on filtrate tubing)
Tygon® tubing (for filtrate; E-3603)
polyethylene tubing (pressure)
polysulfone stir bar(s)
silicone O-ring
silicone gasket

Quality Level

product line

Amicon®

manufacturer/tradename

Amicon®

parameter

50 mL process volume

technique(s)

protein clean-up: suitable (desalting)
protein purification: suitable (concentration)

H

11.7 cm (4.6 in.)

filter diam.

44.5 mm

filtration area

13.4 cm2

shipped in

ambient

Application

This Amicon stirred cell model provides high flow rates with solutions up to 10% macrosolute concentration. Capable of rapid concentration, or salt removal followed by concentration in the same unit.

Principle

Pressure

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
Tygon is a registered trademark of Saint-Gobain Performance Plastics Corp.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Amicon Stirred Cells concentrate and buffer-exchange macrosolutes efficiently with pressure-driven filtration.

Protocols

User guide for Amicon® Stirred cells including features, operation, specifications, and troubleshooting.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service