Skip to Content
Merck
All Photos(1)

Key Documents

SX0995

Supelco

Stoddard Solvent

Synonym(s):

Stoddard Solvent, Mineral Spirits

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116121
EC Index Number:
232-489-3
bp:
153-210 °C
Pricing and availability is not currently available.

form

liquid

color

APHA: ≤20

bp

153-210 °C

transition temp

flash point 38 °C

shipped in

ambient

storage temp.

room temp

SMILES string

BrCCCCCCO.OC(=O)C

InChI

1S/C6H13BrO.C2H4O2/c7-5-3-1-2-4-6-8;1-2(3)4/h8H,1-6H2;1H3,(H,3,4)

InChI key

ORSNIUUONGZHEH-UHFFFAOYSA-N

General description

Stoddard Solvent (white spirit/mineral spirit) is a petroleum distillate composed of a complex mixture of linear and branched alkanes (paraffins), cycloalkanes, and aromatic hydrocarbons. It is widely used as a solvent in the paint industry. It is also used as a dry cleaning agent and a solvent in some types of photocopier toners, adhesives, aerosols, asphalt products, printing inks, wood preservatives, lacquers, varnishes, and industrial cleaning and degreasing applications.[1][2][3]

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

Boiling range153-210°CColor (APHA)20 maxFormClear liquid

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 3 - STOT RE 1

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of ?2 u-Globulin Nephropathy and Adrenal Medullary Pheochromocytomas in Male Rats Following Exposure to Stoddard Solvent IIC
Doi Adriana M, et al.
Inhalation Toxicology, 16(5), 247-257 (2004)
Toxic Effects of Solvent Exposure
Hahn Tilman, et al.
Handbook of solvents., 1347-1347 (2019)
Airborne concentrations of benzene and mineral spirits (stoddard solvent) during cleaning of a locomotive generator and traction motor
Madl AK and Paustenbach DJ
Journal of Toxicology and Environmental Health. Part A, 65(23), 1965-1979 (2002)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service