Skip to Content
Merck
All Photos(2)

Key Documents

SCWP14250

Millipore

MCE Membrane Filter, 8.0 μm Pore Size

MF-Millipore, filter diam. 142 mm, hydrophilic

Synonym(s):

MF-Millipore Membrane Filter, 8 µm pore size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24
pore size:
8.0 μm pore size
84 % porosity
material:
mixed cellulose esters (MCE) membrane
plain filter
white filter
feature:
hydrophilic
filter diam.:
142 mm
thickness:
135 μm

material

mixed cellulose esters (MCE) membrane
plain filter
white filter

Quality Level

description

142 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 50 discs

feature

hydrophilic

manufacturer/tradename

MF-Millipore
Millipore

parameter

620 mL/min-cm2 water flow rate
65 L/min-cm2 air flow rate
75 °C max. temp.

filter diam.

142 mm

thickness

135 μm

gravimetric extractables

6%

refractive index

n/D 1.515

matrix

MF-Millipore

pore size

8.0 μm pore size
84 % porosity

capacity

150 μg/cm2 binding capacity (protein)

bubble point

≥0.42 bar, air with water at 23 °C

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Application

  • QC of fluid holding tanks
  • Fluid monitoring
  • Air monitoring
  • Particle collection
  • Particle analysis

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service