SCT262
BioTracker™ Mitochondrial FerroGreen Live Cell Probe
Human
Synonym(s):
mitochondrial iron live cell probe, mitochondrial iron probe
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
BioTracker™ Mitochondrial FerroGreen Live Cell Probe,
biological source
human
Quality Level
packaging
vial of 100 μg
manufacturer/tradename
Millipore
technique(s)
cell culture | stem cell: suitable
flow cytometry: suitable
shipped in
2-8°C
storage temp.
2-8°C
General description
SCT262 is a novel fluorescent probe for the detection of ferrous ion (Fe2+) in mitochondria where Fe-S clusters and heme proteins are synthesized, and enables live cell fluorescent imaging of intracellular Fe2+. The SCT262 probe has no chelating ability. Add 53 uL DMSO to the 50 ug probe in the tube to create 1mM stock solution. Dilute with HBSS to prepare 5uM working solution.Cells may be incubated for 30 mins in the working solution, after which the supernatant solution can be replaced with the desired treatment solution.SPECTRAL PROPERTIESλex: 505 nmλem: 535 nmSuitable for use with FITC/GFP laser or filter
Features and Benefits
This probe is designed for the detection of mitochondrial ferrous iron in live cells by fluorescent microscopy or flow cytometry.
Target description
Iron is thought to be the most abundant transition metal element in many organisms, participating in diverse physiological activities. Recently, free iron in living cells has received attention due to its significance related to cellular damage or death. Free iron exists in its stable redox states, ferrous ion (Fe2+) and ferric ion (Fe3+). In living cells, understanding the behavior of Fe2+ is important because of the intracellular reductive environment, metal porters and water solubility of Fe2+. SCT262 is a novel fluorescent probe for the detection of ferrous ion (Fe2+) in mitochondria where Fe-S clusters and heme proteins are synthesized. This probe enables live cell fluorescent imaging of intracellular Fe2+. SCT262 has no no chelating ability. Mitochondrial FerroGreen and Fe2+ react irreversibly, which is different from the detection principle of calcium-iron probes such as Fluo-3.
Storage and Stability
Store BioTracker™ Mitochondrial FerroGreen Live Cell Probe at -20 °C, protected from light.
Other Notes
Live cell probe for fluorescent detection of ferrous iron (Fe2+)in mitochondria by fluorescence microscopy or flow cytometry. SCT262 is optimized for the imaging of intracellular iron in live cell applications. IMPORTANT APPLICATION NOTE: Dissolve the lyophilized probe in in DMSO to make a 1 mM stock solution, and dilute with HBSS or serum-free medium to make a 5 uM immediately before staining. Use DMSO-based stock solution same day. Alternatively, probe dissolved in ethanol is stable at -20C for two weeks.
Legal Information
BioTracker is a trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service