Skip to Content
Merck
All Photos(1)

Documents

SCT057

Sigma-Aldrich

BioTracker ERthermAC Temperature-Sensitive Live Cell Dye

Synonym(s):

Live cell imaging probe

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171501
NACRES:
NA.32

technique(s)

cell based assay: suitable

General description

Brown adipocyte tissue (BAT) has been shown to be present in human newborns, small infants, as well as adults. Stimulation of BAT energy expenditure is considered a potential new drug target for obesity and diabetes treatments, and there is increasing interest in the discovery of novel mechanisms and substances for this purpose. However, to facilitate the development of breakthrough therapies, it is essential that techniques to measure the signature function of BAT are readily available.

The BioTracker ERthermAC thermosensitive live cell dye is a novel BODIPY-based dye that is capable of imaging heat production in individual brown adipocytes. The dye′s features such as small size, excellent cellular uptake, low cytotoxicity and robust photostability make it very biocompatible. Data obtained using this dye is comparable with that obtained by conventional methods measuring mitochondrial depolarization, oxygen consumption, and extracellular acidification rates, and with thermal power measurements obtained using the calScreener.

Spectral Properties:
  • Absorption: 565 nm
  • Emissions: 590 nm (Orange/Red) in polarized mitochondria
  • Emissions: 527 nm (Green) in depolarized mitochondria


References:
Kriszt R et al. Optical visualization of thermogenesis in stimulated single-cell brown adipocytes. Sci Rep. 2017 May 3;7(1):1383.

Application

Application: Live cell fluorescent imaging
Live cell imaging thermosensitive dye to visualize heat production in brown adipocytes.

Quality

Purity: ≥98% confirmed by HNMR, LC-MS and HPLC and elemental analysis

Molar Mass: 541.28 g/mol

Other Notes

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Legal Information

BioTracker is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service