Skip to Content
Merck
All Photos(1)

Key Documents

SCC157

Sigma-Aldrich

6CFSMEo- Human Cystic Fibrosis Submucosal Gland Epithelial Cell Line

Human

Synonym(s):

6CFSMEO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81
Pricing and availability is not currently available.

Product Name

6CFSMEo- Human Cystic Fibrosis Submucosal Gland Epithelial Cell Line, 6CFSMEo- human CF submucosal gland epithelial cell line was derived from a cystic fibrosis patient who was compound heterozygote for the ΔF508 and Q2X CFTR mutations.

biological source

human

technique(s)

cell based assay: suitable
cell culture | mammalian: suitable

shipped in

ambient

General description

Cystic fibrosis (CF) is a lethal autosomal recessive disease caused by mutations in the CF transmembrane conductance regulator (CFTR) gene which functions as a cAMP-activated and phosphorylated-regulated Cl channel. In CF, altered Cl- transport and secretion results in the production of thick and viscous mucus that can damage many of the body’s organs. Tracheobronchial submucosal glands secrete mucins and antimicrobial substances that keep the airways sterile along with fluids that help hydrate airway surfaces. The relationship between CF and mucus secretion is unclear and require further investigations [1] [2].

6CFSMEo- is a human tracheobronchial submucosal gland epithelial cell line isolated from an individual with CF, who was compound heterozygote for the ΔF508 and Q2X CFTR mutations [1] [2]. ΔF508 mutation is a trinucleotide deletion that results in loss of a phenylalanine at amino acids 508 (ΔF508) in the CFTR protein. This mutation accounts for ~66% of all CF alleles [1]. Q2X mutation is a rare CF mutation in exon 1 of the CFTR gene in which the second codon (CAG) is mutated into the stop codon UAG. The 6CFSMEo- cell line was immortalized with the origin-of-replication defective SV40 plasmid (pSVori-) [1] [2].

6CFSMEo- retains the characteristic cobblestone morphology of epithelial cells along with cytokeratin expression and the ability to form tight junctions. The cell line expresses vestigial amounts of CFTR mRNA transcripts but does not express detectable levels of CFTR protein [2]. 6CFSMEo- lacks cAMP-induced Cl- currents [2].

Cell Line Description

Epithelial Cells

Application

6CFSMEo- human CF submucosal gland epithelial cell line was derived from a cystic fibrosis patient who was compound heterozygote for the ΔF508 and Q2X CFTR mutations.
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact [email protected].

Quality

• Each vial contains ≥ 1X10⁶ viable cells.
• Cells are tested by PCR and are negative for HPV-16, HPV-18, Hepatitis A, C, and HIV-1 & 2 viruses as assessed by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
• Cells are negative for mycoplasma contamination.
• Each lot of cells is genotyped by STR analysis to verify the unique identity of the cell line.

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Disclaimer

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges {HCompany} to request certain information about you and the establishment where the GMOs are being handled. Click here for Enduser Declaration (EUD) Form.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Carina da Paula et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 15(6), 251-262 (2005-07-23)
Cultured airway epithelial cells are widely used in cystic fibrosis (CF) research as in vitro models that mimic the in vivo manifestations of the disease and help to define a specific cellular phenotype. Recently, a number of in vitro studies
A L Cozens et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(11), 5171-5175 (1992-06-01)
Tracheobronchial glands were isolated and cultured from a patient with cystic fibrosis (CF). Cultured epithelial cells were transformed with pSVori-. All transformed cell lines express cytokeratin filaments and at early passages express the junctional complex molecule cell CAM 120/80, indicating

Articles

16HBE14o- human bronchial epithelial cells used to model respiratory epithelium for the research of cystic fibrosis, viral pulmonary pathology (SARS-CoV), asthma, COPD, effects of smoking and air pollution. See over 5k publications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service