Skip to Content
Merck
All Photos(1)

Documents

QIA91

Sigma-Aldrich

Caspase-3 Detection Kit (FITC-DEVD-FMK)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

usage

sufficient for 100 tests

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

technique(s)

flow cytometry: suitable

input

sample type cultured cells

detection method

fluorometric

shipped in

dry ice

storage temp.

−20°C

General description

A convenient and sensitive method for the detection of activated caspase-3 in living cells. The fluorescent marker, FITC-DEVD-FMK, is a cell-permeable, non-toxic inhibitor that binds irreversibly to activated caspase-3 in apoptotic cells. The fluorescence intensity can be measured by fluorescence microscopy, fluorescence plate reader, or flow cytometry.

Specificity

A broad range of species

Packaging

1 kit in Glass bottle

Components

Labeled Caspase-3 Inhibitor (FITC-DEVD-FMK), Wash Buffer, Caspase-3 Inhibitor (Z-VAD-FMK), and a user protocol.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS

Specifications

Assay Time: 1.5 h

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service