Skip to Content
Merck
All Photos(1)

Documents

PX1505

Supelco

Potassium iodide

crystals, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-659-4

Agency

meets USP/NF

Quality Level

vapor pressure

1 mmHg ( 745 °C)

Assay

≥99.0%

form

crystals

impurities

≤0.005% Insoluble matter

loss

≤0.2% loss on drying, 150°C

color

white

pH

6.9 (20 °C, 50 g/L in H2O)

bp

1325 °C/1013 hPa

mp

681 °C (lit.)

anion traces

chloride, bromide (as Cl)-: ≤0.01%
iodate (IO3-): ≤3 ppm
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.002%
Ca: ≤0.002%
Fe: ≤3 ppm
Mg: ≤0.001%
Na: ≤0.005%
heavy metals (as Pb): ≤5 ppm

shipped in

ambient

storage temp.

room temp

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

crystals, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay99.0% minBarium (Ba)0.002% maxCalcium (Ca)0.002% maxChloride and bromide (as Cl)0.01% maxColorWhiteFormCrystals or fine granulesHeavy metals (as Pb)5ppm maxInsoluble matter0.005% maxIodate (IO3)3ppm maxIron (Fe)3ppm maxLoss on drying (150°C)0.2% maxMagnesium (Mg)0.001% maxPhosphate (PO4)0.001% maxSodium (Na)0.005% maxSulfate (SO4)0.005% maxpH of a 5% solution at 25°C6.0-9.2

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service