Skip to Content
Merck
All Photos(1)

Key Documents

PX1405

Supelco

Potassium chloride

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-211-8
grade:
ACS reagent
form:
crystals

grade

ACS reagent

Quality Level

Agency

meets USP/NF

Assay

99.0-100.5% (silver nitrate titration)

form

crystals

impurities

≤0.005% Insoluble matter

color

white

pH

5.5-8.0 (25 °C, 50 g/L in H2O)

bp

1413 °C/1013 hPa

mp

770 °C (lit.)

density

1.98 g/mL at 25 °C (lit.)

anion traces

bromide (Br-): ≤0.01%
chlorate, nitrate (as NO3-): ≤0.003%
iodide (I-): ≤0.002%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.001%

cation traces

Ba: ≤0.001%, passes test
Ca: ≤0.002%
Fe: ≤3 ppm
Mg: ≤0.001%
Na: ≤0.005%
heavy metals (as Pb): ≤5 ppm

shipped in

ambient

storage temp.

room temp

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay (argentometric)99.0-100.5%Barium (Ba)Passes test (Limit about 0.001%)Bromide (Br)0.01% maxCalcium (Ca)0.002% maxChlorate and nitrate (as NO3)0.003% maxColorWhiteFormCrystalsHeavy metals (as Pb)5ppm maxInsoluble matter0.005% maxIodide (I)0.002% maxIron (Fe)3ppm maxMagnesium (Mg)0.001% maxPhosphate (PO4)5ppm maxSodium (Na)0.005% maxSulfate (SO4)0.001% maxpH of a 5% solution at 25°C5.4-8.6

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service