Skip to Content
Merck
All Photos(1)

Key Documents

PX1130

Sigma-Aldrich

3-Picoline

Synonym(s):

3-Picoline, 3-Methylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
93.13
MDL number:
UNSPSC Code:
12352100
EC Index Number:
203-636-9
Pricing and availability is not currently available.

vapor pressure

6 hPa

Quality Level

Assay

≥98% (GC)

form

liquid

pH

10 (20 °C, 100 g/L in H2O)

bp

144 °C/1013 hPa

mp

-18 °C

transition temp

flash point 40 °C

shipped in

ambient

storage temp.

room temp

SMILES string

n1cc(ccc1)C

InChI

1S/C6H7N/c1-6-3-2-4-7-5-6/h2-5H,1H3

InChI key

ITQTTZVARXURQS-UHFFFAOYSA-N

Analysis Note

Assay (GC): 98% min
Color: Yellow
Form: Clear liquid
Identity (IR-spectrum): Conforms
Water (H2O): 400ppm max

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

98.6 °F - closed cup

Flash Point(C)

37 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service