Skip to Content
Merck
All Photos(1)

Documents

PX0101

Sigma-Aldrich

1-Pentanesulfonic Acid, Sodium Salt, Monohydrate

suitable for HPLC

Synonym(s):

1-Pentanesulfonic Acid, Sodium Salt, Monohydrate, Pentane Sulfonic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4SO3Na · H2O
CAS Number:
Colour Index Number:
45410
MDL number:
UNSPSC Code:
12352107

form

crystalline solid

technique(s)

HPLC: suitable

shipped in

ambient

storage temp.

room temp

InChI

1S/C5H12O3S/c1-2-3-4-5-9(6,7)8/h2-5H2,1H3,(H,6,7,8)/p-1

InChI key

RJQRCOMHVBLQIH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

HPLC

Application

1-Pentanesulfonic Acid, Sodium Salt, Monohydrate can be used as an ion-pairing reagent in HPLC for the analysis of various organic compounds, metabolites, peptides, and proteins.

Analysis Note

Color: White
Form: Crystalline solid
Purity: 99% min
Solubility in water (5% w/v): To pass test
UV Abs. at 210 nm: 1.00AU max
UV Abs. at 220 nm: 0.10AU max
UV Abs. at 254 nm: 0.06AU max
UV Abs. at 280 nm: 0.06AU max

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fuad Al-Rimawi et al.
Journal of chromatography. A, 1160(1-2), 326-335 (2007-06-05)
The potential of methacrylate-based mixed-mode monolithic stationary phases bearing sulfonic acid groups for the separation of positively charged analytes (alkylanilines, amino acids, and peptides) by capillary electrochromatography (CEC) is investigated. The retention mechanism of protonated alkylanilines as positively charged model

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service