PRCYTMAG40K-PX24
MILLIPLEX® Non-Human Primate Cytokine/Chemokine Panel II - 24-Plex Premixed Immunology Multiplex Assay
Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in non-human primate serum, plasma and cell culture samples.
About This Item
Quality Level
species reactivity
nonhuman primates
manufacturer/tradename
Milliplex®
assay range
standard curve range: 1-1,500 pg/mL
(IL-17A)
standard curve range: 10-10,000 pg/mL
(sCD137, IL-2, IP-10/CXCL10)
standard curve range: 146-150,000 pg/mL
(IL-4)
standard curve range: 15-15,000 pg/mL
(IL-21, IL-31)
standard curve range: 20-20,000 pg/mL
(FGF-2, IL-33)
standard curve range: 24-25,000 pg/mL
(IL-23)
standard curve range: 29-30,000 pg/mL
(IL-1α, IL-28A)
standard curve range: 293-300,000 pg/mL
(IL-17E/IL-25)
standard curve range: 3-3,000 pg/mL
(Eotaxin/CCL11, IL-6)
standard curve range: 4-4,000 pg/mL
(Granzyme B)
standard curve range: 7-7,000 pg/mL
(MIP-3α/CCL20)
standard curve range: 73-75,000 pg/mL
(IL-16)
standard curve range: 78-80,000 pg/mL
(Granzyme A)
standard curve range: 8-8,000 pg/mL
(sFasL)
standard curve range: 98-100,000 pg/mL
(Fractalkine/CX3CL1, IL-22, Perforin, TNFβ/LTA)
technique(s)
multiplexing: suitable
compatibility
configured for Premixed
detection method
fluorometric (Luminex xMAP)
shipped in
wet ice
General description
The MILLIPLEX® Non-Human Primate Premixed Cytokine Panel II is a 25 plex kit to be used for the simultaneous quantification of the following analytes in monkey serum, plasma, tissue/cell lysate and culture supernatant samples: sCD137, Eotaxin, sFASL, FGF-2, Fractalkine, Granzyme A, Granzyme B, IL-1α, IL-2, IL-4, IL-6, IL-16, IL-17A, IL-17E/IL-25, IL-21, IL-22, IL-23, IL-28A, IL-31, IL-33, IP-10, MIP-3α, Perforin, RANTES and TNFβ.
The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.
Panel Type: Cytokines/Chemokines
Specificity
This multiplexed assay panel has been tested with samples from multiple non-human primate species. Please see protocol for details.
Application
- Analytes: sCD137, Eotaxin/CCL11, sFasL, FGF-2, Fractalkine/CX3CL1, Granzyme A, Granzyme B, IL-1α, IL-2, IL-4, IL-6, IL-16, IL-17A, IL-17E/IL-25, IL-21, IL-22, IL-23, IL-28A, IL-31, IL-33, IP-10/CXCL10, MIP-3α/CCL20, Perforin, TNFβ/LTA
- Recommended Sample type: serum, plasma and cell culture samples
- Recommended Sample dilution: 1:2 diluted plasma or serum
- Assay Run Time: Overnight or one day. For best results, an overnight incubation is recommended
- Research Category: Inflammation & Immunology
Components
Non-Human Primate Cytokine Panel 2 Quality Control 1, 1 vial, lyophilized
Non-Human Primate Cytokine Panel 2 Quality Control 2, 1 vial, lyophilized
Serum Matrix (0.08% Sodium Azide), 1 bottle, lyophilized
Sample Plate, 96 well format, 1 plate
2 Foil plate sealers
Assay Buffer, 2 bottles, 30 mL
Bead Diluent, 1 bottle, 3.5 mL
Wash Buffer, 10X (0.05% Proclin), 2 bottlesl, 30 mL
Non-Human Primate Cytokine Panel 2 Detection Antibodies, 1 bottle, 3.2 mL
Streptavidin-Phycoerythrin, 1 bottle, 3.2 mL
Premixed 24-plex Beads, 1 bottle, 3.5 mL
Storage and Stability
Other Notes
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2
Target Organs
Respiratory Tract
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service