Skip to Content
Merck
All Photos(1)

Key Documents

PLBC15005

Millipore

Regenerated Cellulose Membrane Filter, 3 kDa NMWCO

Ultracel®, filter diam. 150 mm

Synonym(s):

Ultrafiltration Discs, 3 kDa NMW

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
eCl@ss:
32031602
NACRES:
NB.24
Pricing and availability is not currently available.

material

regenerated cellulose membrane

Quality Level

feature

hydrophilic

manufacturer/tradename

Ultracel®

technique(s)

protein clean-up: suitable (desalting)
protein purification: suitable (concentration)

diam.

150 mm

filter diam.

150 mm

filtration area

177 cm2

matrix

Ultracel®

pore size

3 kDa NMWCO

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Assemble your Amicon Stirred Cell with the membrane of your choice. Ultrafiltration cut disc membranes are available in Ultracel regenerated cellulose and Biomax polyethersulvone (PES). The membranes are available in a range of different pore sizes & diameters.
Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.
Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.
Microfiltration Durapore PVDF cut discs are available 0.1 µm, 0.22 µm, and 0.45 µm pore sizes in both 63.5 mm and 75 mm diameters.

Features & Benefits:
•Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm
•Both Ultracel, Durapore, and Biomax membranes are available in a wide range of pore sizes to meet your separation requirements
•Compatible with Amicon Stirred Cells

Applications:
Concentration, Buffer Exchange, Desalting
Device Configuration: Ultrafiltration Discs
Filter Code: PLBC
Filter Type: Ultrafiltration

Application

3 kDa NMWL, Ultracel regenerated cellulose, 150 mm diameter

Legal Information

ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service