Skip to Content
Merck
All Photos(1)

Key Documents

PC635

Sigma-Aldrich

PhosphoDetect Anti-NBS1 (pSer³⁴³) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Nibrin (pSer³⁴³) Rabbit pAb

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

diluted serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer343)

Gene Information

human ... NBN(4683)

General description

Rabbit polyclonal antibody supplied as adsorbed diluted serum. Recognizes the ~95 kDa nibrin (NBS1) protein phosphorylated at Ser343.
Recognizes the ~95 kDa nibrin protein phosphorylated at Ser343 in δ-irradiated lymphoblastoid cells.
This PhosphoDetect Anti-NBS1 (pSer³⁴³) Rabbit pAb is validated for use in ELISA, Immunoblotting, Immunoprecipitation for the detection of NBS1 (pSer³⁴³).

Immunogen

Human
a synthetic phosphopeptide [TPGPSLpSQGVSVD(C)] corresponding to amino acids 337-349 surrounding the Ser³⁴³ phosphorylation site of human nibrin, conjugated to diphtheria toxoid

Application

ELISA (1:16,000)

Immunoblotting (1:1000)

Immunoprecipitation (1 µl/500 µg cell extract)

Warning

Toxicity: Standard Handling (A)

Physical form

Serum diluted in PBS.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
γ-irradiated lymphoblastoid cells

Other Notes

Gatei, M., et al. 2000. Nat. Genet.25, 115.
Lim, D.S., et al. 2000. Nature404, 613.
Wu, G., et al. 2000. J. Biol. Chem.275, 30618.
Zhong, Q., et al. 1999. Science285, 747.
Carney, J.P., et al. 1998. Cell93, 477.
Trujillo, K.M., et al. 1998. J. Biol. Chem.273, 21447.
Varon, R., et al. 1998. Cell93, 467.
This antibody preparation has been adsorbed against non-phosphopeptide that corresponds to the immunogen sequence. Does not detect non-phosphorylated nibrin protein. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service