Skip to Content
Merck
All Photos(1)

Key Documents

MXRPCLKT1

Millipore

Milliflex® Rapid Cleaning Kit

For 20 cleanings and 2 decontaminations

Synonym(s):

Decontamination Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116130
NACRES:
NB.79
Pricing and availability is not currently available.

packaging

pkg of 1 kit

manufacturer/tradename

Milliflex®

technique(s)

Rapid Sterility Testing: suitable

application(s)

bioburden testing
environmental
food and beverages
pharmaceutical
sterility testing

compatibility

Milliflex® Rapid

storage temp.

room temp

General description

Milliflex® Rapid Cleaning Kit is a unit of the Milliflex® Rapid Microbiology Detection System, which is an automated solution for the rapid detection, response, and resolution of microbial contamination in filterable samples throughout the manufacturing process. The AutoSpray Station issues a warning message across the control panel when calibration, cleaning, or priming are required ensuring consistent and reliable performance throughout the life of the system. Milliflex® Rapid Cleaning Kit is a self-cleaning unit, and the AutoSpray Station will be ready for re-use in five minutes. The unit should be recalibrated after every 5000 samples, or at least once a year.

Application

Milliflex® Rapid Cleaning Kit is a self-cleaning unit to be used with the Milliflex® Rapid Microbiology Detection System.

Components

  • 4 x rinsing agent vials
  • 4 x cleaning agent vials
  • 1 x decontamination vial
  • 10 x Luer-lok® caps
  • 9 x Vials adapters
  • 1 x user guide

Legal Information

Luer-Lok is a registered trademark of Becton-Dickinson & Co.
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Milliflex® Rapid System 2.0 accelerates microbial detection in quality control labs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service