Skip to Content
Merck
All Photos(1)

Documents

MVFCN1225

Millipore

Multiscreen® 96 well Plate, glass fiber membrane

pore size 1.2 μm, non-sterile

Synonym(s):

96-Well Glass Fiber Plate, Glass Fiber 96-Well Plate, MultiScreen Glass Plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NB.22

material

flat bottom wells
glass fiber membrane
polypropylene

Quality Level

description

Opaque, Non-Sterile, with Glass Fiber membrane, 1.2 µm pore size, 25 plates, Polypropylene device

sterility

non-sterile

product line

MultiScreen®

feature

lid: no

manufacturer/tradename

MultiScreen®

parameter

2 mL sample volume (per well)

technique(s)

ligand binding assay: suitable
protein purification: suitable (TCA precipitation)
receptor binding assay: suitable

H

47.2 mm

L

127.8 mm

W

85.6 mm

filtration area

0.23 cm2

plate size

96 wells

well maximum volume

2 mL

working volume

0.1-2 mL

pore size

1.2 μm pore size

application(s)

sample preparation

binding type

high binding surface

shipped in

ambient

General description

  • MultiScreen® 96 well high volume filter plates with 1.2 μm Glass Fiber C (GFC) membrane offers superior vacuum filtration and filtrate collection.
  • Plates are chemically resistant to both aqueous and organic solutions.
  • Uniquely designed and are fully automation-compatible.

Application

  • Lysate clearing
  • Viscous sample filtration
  • Environmental testing
  • DNA sample preparation bind-wash-elute applications

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

ELISpot assay provides qualitative and quantitative information on immune responses, visualizing multiple secretory products from single responding cells.

Related Content

This provides information about the chemical compatibility of MultiScreen Filter Plates with different reagents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service