Select a Size
About This Item
Skip To
form
liquid
Related Categories
1 of 4
This Item | MP8W15 | MP10W15 | MP10W10 |
|---|---|---|---|
| form liquid | form - | form - | form - |
Application
Do not boil.
Western Blotting
General description
- Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
10 - Combustible liquids
wgk
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.
Refresh your gel casting workflow with ready-to-use SDS-PAGE gels in just 3 minutes. Discover faster, simpler, and safer gel casting methods.
Protocols
Visão geral da química de géis de poliacrilamida e instruções detalhadas para moldagem manual de géis de poliacrilamida com o uso dos kits de moldagem de géis bis-tris mPAGE® TurboMix.
Simplify your SDS-PAGE gel preparation with our pre-mixed solutions and protocols to achieve precise protein separation.
Related Content
Nós entendemos. Os faroestes podem ser uma dor no borrão, e nós certamente já fizemos a nossa parte. Portanto, compartilhe conosco seus borrões feios e receba uma surpresa! E, embora não possamos dizer o que é - isso estragaria a surpresa -, exceto que isso despertará a criatividade no laboratório.
We get it. Westerns can be a pain in the blot, and we’ve certainly made our share. So, share with us your ugly blots and receive a surprise! And while we can’t tell you what it is – that would spoil the surprise - except that it will spark creativity in the lab.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


