MP42G10
mPAGE® 4-20% Bis-Tris Precast Gels
4-20% Bis-Tris, 10 well, 1mm
Synonym(s):
Bis-Tris precast gels, precast gels for SDS-PAGE, precast polyacrylamide gels
About This Item
Recommended Products
product name
mPAGE® 4-20% Bis-Tris Precast Gel, L × W × wells 10 cm × 8 cm × 10 well
shelf life
18 mo.
packaging
pkg of 10 gels
L × W × wells
10 cm × 8 cm × 10 well
cassette W × L
10 cm × 8 cm
gel thickness
1 mm
well maximum volume
80 μL
well wells
10 wells
compatibility
mPAGE® MES or MOPS Running Buffer; mPAGE® Mini Gel System
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Application
- Load up to 80 μl sample / well
- Typical run time 30 min (MES, 180V) or 34 min (MOPS, 200V)
- Reverse gasket when using with Bio-Rad Mini-PROTEAN systems
- Use adapter (MPTA) with XCell SureLock and other compatible systems
Western Blotting
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Bis-Tris SDS-PAGE gel casting kits with mPAGE® TurboMix technology for rapid, reliable gel casting and protein separation.
Related Content
mPAGE® gels offer high-resolution protein separation, reduced cost, and compatibility with common electrophoresis tanks.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service