Skip to Content
Merck
All Photos(5)

Key Documents

MMHEADEZ1

Millipore

Milliflex Oasis® Filtration Head

for use with EZ-Fit® filtration units

Synonym(s):

Milliflex Oasis® Head, Milliflex Oasis® filtration pump head

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.79
Pricing and availability is not currently available.

material

316L stainless steel (ASI)

packaging

pkg of 1 unit

manufacturer/tradename

Milliflex Oasis®

application(s)

bioburden testing
food and beverages
food and beverages
pharmaceutical
water monitoring

compatibility

for use with EZ-Fit® filtration units

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

The Milliflex Oasis® head for EZ-Fit® filtration unit allows the use of EZ-Fit® filtration units with the Milliflex Oasis® filtration pump. The EZ-Fit® filtration unit is a disposable filtration device for bioburden testing and have been designed to ensure optimal microbial recovery. The sterilized, disposable filtration devices streamline the workflow, save time and give highly reliable microbial enumeration results. The Milliflex Oasis® head enables easy attachment of the filtration unit to the Milliflex Oasis® filtration pump.

Application

Milliflex Oasis® Head is an accessory to be used in Milliflex Oasis® filtration pump and finds its application in bioburden testing and water monitoring of beer, bottled water, cosmetics and Pharmaceutical products analysis.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

configured for

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service