Skip to Content
Merck
All Photos(2)

Key Documents

MBNMAG-41K

Millipore

MILLIPLEX® Mouse Bone Magnetic Bead Panel - Bone Metabolism Multiplex Assay

The analytes available for this multiplex kit are: ACTH, DKK-1, FGF-23, IL-6, Insulin, Leptin, Osteoprotegerin, SOST, TNF-α.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

mouse

manufacturer/tradename

Milliplex®

assay range

accuracy: 84-94%
standard curve range: 10-40,000 pg/mL
(Leptin)

standard curve range: 15-60,000 pg/mL
(DKK-1)

standard curve range: 16-65,000 pg/mL
(OPG)

standard curve range: 3-12,000 pg/mL
(SOST)

standard curve range: 3-12,000 pg/mL
(TNFα)

standard curve range: 37-150,000 pg/mL
(Insulin)

standard curve range: 4-15,000 pg/mL
(ACTH)

standard curve range: 7-30,000 pg/mL
(FGF-23)

standard curve range: 7-30,000 pg/mL
(IL-6)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

Bone metabolism is the dynamic process of ongoing bone deposition and resorption, controlled by osteoblasts, osteocytes, and osteoclasts. While osteoblasts and osteocytes (osteoblasts surrounded by matrix) are responsible for bone deposition, osteoclasts are responsible for bone resorption. Both are required to maintain bone structure, as well as an adequate supply of calcium. To maintain this metabolic balance these cells rely on complex signaling pathways involving hormones and cytokines to achieve the appropriate rates of growth and differentiation. The disruption of bone metabolism results in such disease as osteoporosis, osteoarthritis, rheumatoid arthritis, chronic kidney disease and bone metastases.

MILLIPLEX® Mouse Bone Panel is a 9-plex kit to be used for the simultaneous quantification of any or all of the following analytes in serum, plasma and tissue culture supernatant samples: ACTH, IL-6, OPG, Insulin, Leptin, DKK1, SOST, TNFα and FGF23. This kit uses a 96-well format, contains a lyophilized standard cocktail, two internal assay quality controls and can measure up to 38 samples in duplicate.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Bone

Application

  • Analytes: ACTH, DKK-1, FGF-23, IL-6, Insulin, Leptin, Osteoprotegerin (OPG), Sclerostin (SOST), TNF-α
  • Recommended Sample Type: Mouse Serum, plasma, cell/tissue culture supernatants or lysates
  • Recommended Sample Dilution: 25 μL per well of 1:2 diluted serum or plasma; cell/tissue culture samples may require dilution in appropriate control medium
  • Assay Run Time: Overnight (16-18 hours) at 2-8°C or 2 hours at room temperature (20-25°C)
  • Research Category: Bone
  • Research Subcategory: Metabolism

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Other Notes

Please contact Technical Service for linearity of dilution.
Sensitivity: See kit protocol for individual analytes sensitivities.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason Plummer et al.
Journal of cellular biochemistry, 118(1), 31-42 (2016-05-19)
Dietary methionine restriction (MR) increases longevity and improves healthspan in rodent models. Young male C57BL/6J mice were placed on MR to assess effects on bone structure and formation. Mice were fed diets containing 0.86% or 0.12% methionine for 5 weeks.
Positive environmental modification of depressive phenotype and abnormal hypothalamic-pituitary-adrenal axis activity in female C57BL/6J mice during abstinence from chronic ethanol consumption.
Pang, TY; Du, X; Catchlove, WA; Renoir, T; Lawrence, AJ; Hannan, AJ
Frontiers in Pharmacology null
Amadou Ouattara et al.
Bone reports, 5, 33-42 (2016-01-01)
Methionine restriction (MR) extends the lifespan of a wide variety of species, including rodents, drosophila, nematodes, and yeasts. MR has also been demonstrated to affect the overall growth of mice and rats. The objective of this study was to evaluate
Dextromethorphan inhibits osteoclast differentiation by suppressing RANKL-induced nuclear factor-?B activation.
Wu, K; Lin, TH; Liou, HC; Lu, DH; Chen, YR; Fu, WM; Yang, RS
Osteoporosis International : A Journal Established as Result of Cooperation Between the European Foundation for Osteoporosis and the National Osteoporosis Foundation of the USA null
Heather A Himburg et al.
Radiation research, 186(2), 141-152 (2016-07-09)
Ionizing radiation exposure can cause acute radiation sickness (ARS) by damaging the hematopoietic compartment. Radiation damages quiescent hematopoietic stem cells (HSCs) and proliferating hematopoietic cells, resulting in neutropenia, thrombocytopenia and increased risk for long-term hematopoietic dysfunction and myelodysplasia. While some

Related Content

UnEnhance research with MILLIPLEX® multiplexing tips, industry guidance, and one-day assays for limited lab time.

Multiplex immunoassays save time and sample volume in metabolic syndrome research, providing comprehensive insights into related conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service