Skip to Content
Merck
All Photos(1)

Documents

MAWP037P0

Millipore

Fluid Contamination Monitor, 37 mm, mixed cellulose esters, 0.8 µm, white, with pad

Colorimetric particle monitoring

Synonym(s):

Fluid monitors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
eCl@ss:
32161002
NACRES:
NB.24

material

cellulose (thick pad)
clear Tenite plastic (housing)
mixed cellulose esters (MCE) membrane
polyethylene (plugs)

sterility

non-sterile

filter diam.

37 mm

filtration area

9 cm2

pore size

0.8 μm pore size

fitting

female Luer inlet slip
female Luer outlet slip

application(s)

industrial qc

shipped in

ambient

General description

Fluid contamination analysis monitors are transparent, disposable, Tenite plastic filter holders, pre-assembled with mixed esters of cellulose membranes and absorbent cellulosic pads. The filter is sealed between the monitor sections with a thick cellulose pad under the filter to distribute sample flow evenly over the filter surface.

Monitors have average background particle count marked on each package. Special thick pad supports the filter at high differential pressures.

For match weight fluid monitors, please see MAWP037PM.

Components (top to bottom):
  • Blue plug (polyethylene)
  • Monitor top (Tenite)
  • 0.8μm MF-Millipore filter, 37 mm (mixed cellulose esters)
  • Thick absorbent pad, 34 mm (cellulose)
  • Monitor bottom (Tenite)
  • Red plug (polyethylene)

Replacement parts
  • AAWP037P0 | MF-Millipore Membrane Filter, 0.8 μm pore size (Monitor refills with thick absorbent pads for liquid monitoring)
  • AP30034P0 | Thick Absorbent Pad, 34 mm
  • M000037P0 | Fluid Contamination Monitor, empty

Principle

Vacuum filtration

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany
Tenite is a trademark of Eastman Chemical Company

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service