Skip to Content
Merck
All Photos(1)

Documents

MABS2233

Sigma-Aldrich

Anti-Intestinal Villus Antibody, clone B6A6

Synonym(s):

Intestinal crypt cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

B6A6, monoclonal

purified by

using protein G

species reactivity

mouse

packaging

antibody small pack of 100 μg

technique(s)

flow cytometry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG2aκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Related Categories

General description

The small intestine consists of continuous villi and crypts. The crypt is mainly a proliferative compartment and is maintained by multipotent stem cells whereas the villus represents the differentiated compartment, and it receives cells from multiple crypts. The functional compartment of the epithelium contains differentiated cells that populate the villi and can be categorized based on their function: enterocytes that function to absorb nutrients, goblet cells that secrete a protective mucus barrier, and enteroendocrine cells that release gastrointestinal hormones. Intestinal crypts contain intestinal stem cells (ISC), which are multipotent adult stem cells that reside in the base of crypts. They continuously self-renew by dividing and differentiate into the specialized cells of the intestinal epithelium, which renews throughout life. ISCs located either at the crypt base interspersed between the Paneth cells can be categorized into active-cycling (Lgr5+ cells) or those located near the þ4 position in the crypt as slow-cycling Bmi-1+ cells. Paneth cells that are responsible for secreting antibacterial peptides reside at the base of the proliferative compartment. As ISCs begin to differentiate, they migrate toward the lumen and are eventually shed, either from the tip of the intestinal villi or from the surface of the colonic epithelium. Villus represents the differentiated compartment of intestinal cells. Clone B6A6 recognizes most villous cells except goblet cells. (Ref.: Smith, NR., et al. (2018). Cell. Mol. Gastroenterol. Hepatol. 6(1);79-96; Wang, F., et al. (2013). Gastroenterology. 145(2); 383-395; Umar, S. (2010). Curr. Gastroenterol. Rep. 12(5); 340-348).

Specificity

Clone B6A6 is a rat monoclonal antibody that detects intestinal villus cells.

Immunogen

Undifferentiated Mouse intestinal epithelial cells. Rats were preimmunized with isolations of differentiated mouse intestinal epithelial cells and then treated with cyclophosphamide (i.p.) to eliminate B lymphocytes reacting against these antigens. Subsequent immunization was with crypt-based single cells.

Application

Quality Control Testing

Isotype testing: Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG2a .

Tested Applications

Flow Cytometry Analysis: A representative lot detected Intestinal Villus in Lgr5-GFP mouse intestinal cells ( Data courtesy of Prof. Melissa Wong, Ph.D., Oregon Health & Science University, Portland, Oregon USA).

Immunohistochemistry Applications: A representative lot detected Intestinal Villus cells in Immunohistochemistry applications (Wang, F., et al. (2013). Gastroenterology. 145(2): 383-95.e1-21).

Flow Cytometry Analysis: A representative lot detected Intestinal Villus in Flow Cytometry applications (Wang, F., et al. (2013). Gastroenterology. 145(2): 383-95.e1-21; Smith, N.R., et al. (2017). Cell Mol Gastroenterol Hepatol. 3(3): 389-409; Yan, K.S., et al. (2017). Cell Stem Cell. 21(1):7 8-90.e6; Smith, N.R., et al. (2018). Cell Mol Gastroenterol Hepatol. 6(1): 79-96).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Intestinal Villus clone B6A6, Cat. No. MABS2233, is a rat monoclonal antibody that detects Intestinal Villus cells and is tested for use in and Flow Cytometry and Immunohistochemistry.

Physical form

Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service