Skip to Content
Merck
All Photos(1)

Documents

MABS1326

Sigma-Aldrich

Anti-Importin alpha-3/Qip1 Antibody, clone 3D10

clone 3D10, from rat

Synonym(s):

Importin subunit alpha-3, Importin alpha Q1, Karyopherin subunit alpha-4, Qip1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D10, monoclonal

species reactivity

rat, monkey, hamster, bovine, human, mouse

species reactivity (predicted by homology)

canine (based on 100% sequence homology)

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

bovine ... Kpna3(540719)
dog ... Kpna3(476909)
human ... KPNA3(3839)
mouse ... Kpna3(16648)
rat ... Kpna3(361055)
rhesus monkey ... Kpna3(706925)

General description

Importin subunit alpha-3 (UniProt O35343; also known as Importin alpha Q1, Karyopherin subunit alpha-4, Qip1) is encoded by the Kpna4 (also known as Qip1) gene (Gene ID 16649) in murine species. The transport of macromolecules between cytoplasm and nucleus is mediated by importins and exportins and regulated by the small GTPase Ran. Importins bind their cargoes with either nuclear localization signal (NLS) or M9 domain-containing hnRNP proteins in the cytoplasm, while exportins target their substrates via their nuclear export signal (NES) or M9 domain inside the nucleus. Qip1/KPNA4 belongs to the importin- /karyopherin- (KPNA) family of adaptor proteins that interact with both monopartite and bipartite types of classical nuclear localization signals (cNLSs). The seven KPNA family members are divided into the 1 (KPNA1/5/6), 2 (KPNA2/7), and 3 (KPNA3/4) subfamilies based on structural and functional similarities. KPNA members mediate substrate cargo nuclear import by bridging substrate cargo and the Ran-karyopherin- complex. Both KPNA3 and Qip1/KPNA4 are reported to target methyl-CpG-binding protein 2 (MeCp2) bipartite NLS (255-RKAEADPQAIPKKRGRK-271). Qip1/KPNA4 can ineract with both wild-type and MeCp2 R270X mutants found in Rett syndrome (RTT) patients, while KPNA3 binds only wild-type, but not MeCp2 R270X mutants. Human and murine Qip1 contains an NLS (a.a. 43-52) in its importin- -binding (IBB) domain (a.a. 2-58), followed by ten ARM domains (a.a. 66-485), one majore and one minor NLS-binding site (a.a. 137-229 and a.a. 306-394).

Specificity

Clone 3D10 detected recombinant mouse importin 3/Qip1, but not importin 1/Rch1, importin 5/NPI-1, or importin 7/S2 GST fusion proteins (Sakaguchi, N., et al. (2003). Hybrid. Hybridomics. 22(6):397-400).

Immunogen

Recombinant full-length mouse Importin alpha-3/Qip1.

Application

Detect Importin subunit alpha-3/Qip1 using this rat monoclonal Anti-Importin alpha-3/Qip1, clone 3D10, Cat. No. MABS1326, validated for use in Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A representative lot detected recombinant mouse importin 3/Qip1, but not importin 1/Rch1, importin 5/NPI-1, or importin 7/S2 GST fusion proteins (Sakaguchi, N., et al. (2003). Hybrid. Hybridomics. 22(6):397-400).

Western Blotting Analysis: A representative lot detected endogenous importin 3/Qip1 in human HeLa, monkey COS-7, mouse L-929, rat NRK , hamster BHK, and bovine MDBK cell lysates (Sakaguchi, N., et al. (2003). Hybrid. Hybridomics. 22(6):397-400).

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected in 10 µg of HeLa cell lysate.

Target description

~58 kDa observed. 57.76 kDa (human) and 57.79 kDa (mouse & rat) calculated (Met1 removed). Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified rat IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zheng Liu et al.
Aging, 15(16), 8487-8500 (2023-08-31)
ZBTB34 is a novel zinc finger protein with an unknown function. In this study, the gene expression and survival prognosis of ZBTB34 were analyzed across tumors based on the TCGA datasets. According to the bioinformatics analysis and qPCR results, liver

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service