Skip to Content
Merck
All Photos(2)

Documents

MABS1241

Sigma-Aldrich

Anti-Synembryn-A/Ric8A Antibody, clone 3E1

clone 3E1, from mouse

Synonym(s):

Protein Ric-8A, Resistance to inhibitors of cholinesterase 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E1, monoclonal

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... RIC8A(60626)

General description

Synembryn-A (UniProt: Q80ZG1; also known as Resistance to inhibitors of cholinesterase 8, Protein Ric-8A) is encoded by the Ric8a gene (Gene ID: 293614) in rat species. Resistance to inhibitors of cholinesterase 8 (RIC-8) is conserved in vertebrates and is concentrated in the cytoplasm of neurons. Mammalian Ric-8 proteins are G alpha subunit guanine nucleotide exchange factors. Ric-8A and Ric-8B collectively stimulate nucleotide exchange of all G alpha subunit classes by stabilizing the G alpha nucleotide-free transition state. Ric-8 is reported to directly participate in the post-translational folding of nascent G alpha subunits to promote G-protein biosynthesis. It may facilitate the transition of G alpha from a pre-folded globular state to its native state by promoting the first G alpha guanine nucleotide-binding event. Ric-8 interacts with GDP-bound G alpha proteins, but does not interact with G-alpha proteins when they are in complex with beta and gamma subunits. Overexpression of Ric-8 proteins in HEK293, NIH 3T3, or Sf9 cells is shown to enhance cellular G alpha levels. Embryonic stem cells lacking Ric-8A or Ric-8B produce defective G alpha subunits that turn over at about 10-fold higher rates than in wild-type cells. Ref.: Thomas, C.J. et al. (2011). PLoS ONE 6(8): e23197.

Specificity

Clone 3E1 detects Ric-8A in human brain.

Immunogen

A full-length recombinant Ric-8A protein from rat.

Application

Detect Synembryn-A/Ric-8A using this mouse monoclonal Anti-Synembryn-A/Ric8A antibody, clone 3E1, Cat. No. MABS1241, validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50-250 dilution from a representative lot detected Synembryn-A/Ric-8A in human cerebral cortex, human cerebellum, and human testis tissues.

Western Blotting Analysis: A representative lot detected recombinant Synembryn-A/Ric-8A protein (Chan, P.Y., et al. (2013). Proc. Natl. Acad. Sci. USA. 110(10):3794-3799).

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Synembryn-A/Ric-8A in 10 µg of human brain tissue lysate.

Target description

~60 kDa observed; 59.7 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service