Skip to Content
Merck
All Photos(2)

Documents

MABS1163

Sigma-Aldrich

Anti-CSF-1R/CD115 Antibody clone FER216

clone FER216, from mouse

Synonym(s):

Macrophage colony-stimulating factor 1 receptor, CD115, CSF-1 receptor, CSF-1-R, CSF-1R, M-CSF-R, Proto-oncogene c-Fms

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

FER216, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CSF1R(1436)

General description

Macrophage colony-stimulating factor 1 receptor (EC 2.7.10.1; UniProt P07333; also known as CD115, CSF-1 receptor, CSF-1-R, CSF-1R, M-CSF-R, Proto-oncogene c-Fms) is encoded by the CSF1R (also known as CSFR, FMS, HDLS) gene (Gene ID 1436) in human. CSF1R is a CSF1/PDGF family receptor tyrosine kinase that mediates macrophage colony stimulating factor 1 (M-CSF1) and IL-34 signaling by acting as the receptor for these two cytokines. CSF1R plays a key role in the differentiation, recruitment and activation of tissue macrophages. Tumor-associated macrophages (TAMs) have been associated with survival in classic Hodgkin lymphoma (cHL) and other lymphoma types. Hyperphosphorylated CSF1R is found in cHL tissue and highlights the importance of CSF1/CSF1R signaling in the recruitment of TAMs. CSF1R is initially produced with a signal peptide sequence (a.a. 1-19), the removal of which yields the mature type I single transmembrane protein with an extracellular region (a.a. 20-517), a transmembrane segment (a.a. a.a. 518-538), and a cytoplasmic region (a.a. 539-972) that contains the kinase domain (a.a. 582-910).

Specificity

Clone FER216 reacts with the extracellular domain of human CSFR1, but not the extracellular domain of human FGFR1. Clone FER216 detected the ~100 kDa human CSFR1 in nomral tonsil and Hodgkin lymphoma (cHL) tissue extracts, as well as an additional ~150 kDa phosphorylated CSFR1 band in cHL tissue extracts (Martín-Moreno, A.M., et al. (2015). PLoS One. 2015 Jun 12;10(6):e0125203).

Immunogen

Epitope: Extracellular domain.
Human IgG1 Fc-tagged recombinant human CSF-1R/CD115 extracellular domain.

Application

Anti-CSF-1R/CD115 Antibody clone FER216 is an antibody against CSF-1R for use in Western Blotting.
Research Category
Signaling
Research Sub Category
Cytokines & Cytokine Receptors
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected CSF-1R/CD115 in 10 µg of human tonsil tissue lysate.

Quality

Evaluated by Western Blotting in human colon tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected CSF-1R/CD115 in 10 µg of human colon tissue lysate.

Target description

~107 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martina Molgora et al.
Cell, 182(4), 886-900 (2020-08-14)
Checkpoint immunotherapy unleashes T cell control of tumors, but is undermined by immunosuppressive myeloid cells. TREM2 is a myeloid receptor that transmits intracellular signals that sustain microglial responses during Alzheimer's disease. TREM2 is also expressed by tumor-infiltrating macrophages. Here, we found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service