biological source
hamster
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
H24G11, monoclonal
mol wt
calculated mol wt 70.33 kDa
observed mol wt ~85 kDa
purified by
using protein G
species reactivity
mouse
packaging
antibody small pack of 100 μL
technique(s)
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable
isotype
IgGκ
epitope sequence
Unknown
Protein ID accession no.
UniProt accession no.
shipped in
2-8°C
target post-translational modification
unmodified
Gene Information
mouse ... Crtac1(72832)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in Mouse E14 brain tissue lysates.
Western Blotting Analysis: A 1:500 dilution of this antibody detected LOTUS in Mouse E14 brain tissue lysates.
Tested Applications
Immunocytochemistry Analysis: A representative lot detected LOTUS in Immunocytochemistry applications (Kurihara, Y., et al. (2014). Mol Cell Neurosci. 61:211-8; Sato, Y., et al. (2011). Science. 333(6043):769-73).
Immunohistochemistry Applications: A 1:50 dilutions from a representative lot detected LOTUS in E14 embryonic mouse tissue sections.
Western Blotting Analysis: A representative lot detected LOTUS in Western Blotting applications (Iketani, M., et al. (2016). Sci Rep. 6:39586).
Immunohistochemistry Applications: A representative lot detected LOTUS in Immunohistochemistry applications ( Sato, Y., et al. (2011). Science. 333(6043):769-73; Iketani, M., et al. (2016). Sci Rep. 6:39586).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service