Skip to Content
Merck
All Photos(1)

Documents

MABN2292

Sigma-Aldrich

Anti-Neuregulin-1 Antibody, clone 4F10

clone 4F10, from rat

Synonym(s):

NRG1, Acetylcholine receptor-inducing activity, ARIA, Breast cancer cell differentiation factor p45, Glial growth factor, Heregulin, HRG, Neu differentiation factor, Sensory and motor neuron-derived factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

antibody form

purified antibody

antibody product type

primary antibodies

clone

4F10, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... NRG1(3084)

General description

Neuregulin-1 (UniProt: Q02297; also known as Acetylcholine receptor-inducing activity, ARIA, Breast cancer cell differentiation factor p45, Glial growth factor, Heregulin, HRG, Neu differentiation factor, Sensory and motor neuron-derived factor) is encoded by the NRG1 (also known as GGF, HGL, HRGA, NDF, SMDF) gene (Gene ID: 3084) in human. Neuregulin-1 is a single-pass type 1 membrane protein that acts as a direct ligand for ErbBB3 and ErbB4 tyrosine kinase receptors. It concomitantly recruits ERBB1 and ERBB2 coreceptors, resulting in ligand-stimulated tyrosine phosphorylation and activation of the ERBB receptors. Eleven isoforms of Neuregulin-1 are known that are produced by alternative splicing. They have been classified as type I (isoforms with an Ig domain and a glycosylation domain) type II NRGs (isoforms with an Ig domain, but no glycosylation domain). These multiple isoforms perform diverse cellular functions, such as inducing growth and differentiation of epithelial, glial, neuronal, and skeletal muscle cells and inducing expression of acetylcholine receptor in synaptic vesicles during the formation of the neuromuscular junction. Proteolytic cleavage close to the plasma membrane on the external face leads to the release of the soluble growth factor form. Dual cleavage of Neuregulin-1 by beta-secretase 1 (BACE1) and ADAM17 is shown to liberate EGF-like domain, which stimulates ErbB3 signaling. (Ref.: Fleck, D., et a. (2013). J. Neurosci. 33(18):7856-7869).

Specificity

Clone 4F10 is a rat monoclonal antibody that detects human Neuregulin-1. It targets an epitope within 10 amino acids from the N-terminal half.

Immunogen

A linear peptide corresponding to 10 amino acids from the N-terminal, extracellular domain of human Neuregulin 1, isoform 6.
Epitope: extracellular domain

Application

Anti-Neuregulin-1 clone 4F10, Cat. No. MABN2292, is a highly specific rat monoclonal antibody that targets Typ III NRG1 beta-1 and has been tested for use in Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A representative lot detected Neuregulin-1 in Western Blotting applications (Fleck, D., et. al. (2013). J Neurosci. 33(18):7856-69; Cheret, C., et. al. (2013). EMBO J. 32(14):2015-28; Fleck, D., et. al. (2016). J Biol Chem. 291(1):318-33; Ben Halima, S., et. al. (2016). Cell Rep. 14(9):2127-2141).

Quality

Evaluated by Western Blotting in HEK293 cells transfected with Type I NRG1 and Type III NRG1 and treated with BACE inhibitor.

Western Blotting Analysis: 1 µg/mL of this antibody detected Neuregulin-1 in HEK293 cells transfected with Type I NRG1 and Type III NRG1 and treated with BACE inhibitor.

Target description

~ 71 kDa observed; 71.16 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service